United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Baths and Circulators
>
Laboratory, Utility Water Baths
> Economical Dual Poly Bath
Economical Dual Poly Bath
Dual chambers to perform two experiments simultaneously
Low-level shut-off protects you and your samples
Corrosion and chemical resistant interior provides years of use
Laboratory, utility water baths are the ideal bath for maintaining a temperature set points over a long period of time. Different size and depth of the bath chamber to meet most every application. Units feature analog or digital temperature control with a ranges from ambient to 100° C. Options of high-temperature and low-level cutoff features to protect samples.
This economical dual-chamber bath provides you the flexibility of performing two different temperature based experiments simultaneously. Separate chambers allow you to set the temperature independently from ambient to 90° C, without temperature interference from the other bath. The dual 5.5 liter chambers allow you to heat samples of varying sizes. Each chamber has its own digital temperature display for easy setting and read-out.
Each chamber interior is made from chemical and corrosion-resistant polypropylene, allowing for easy clean-up and durability. The rugged exterior resists dents, cracking, and chemicals. Low-level sensor automatically shuts the bath down and sounds an alarm if the water level goes below 1-inch.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Dual-Chamber Poly Water Bath, 5.5-liter capacity each chamber, ambient to 90
C, 120VAC
Product Type Water Bath,Bath type Tank Bath,Capacity Liters 5.5
EW-39066-50
Usually ships in 8 days.
$800.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Economical Dual Poly Bath
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Baths and Circulators
>
Laboratory, Utility Water Baths
> Economical Dual Poly Bath
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title