United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$500 to $799
(1)
$800 to $1,099
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Vacuum Pumps
>
Compact/OEM Vacuum Pumps
> Economical Oil Pumps
Economical Oil Pumps
The most economical rotary vane oil pump available.
Centrifugal force holds woven linen vanes against finely polished cast-iron body interior for a vacuum-tight seal—the self-adjusting vanes automatically compensate for wear.
Pumps include separate vacuum and pressure gauges, vacuum and pressure regulators, oil trap, air filter, carrying handle, and a 6-ft, three-wire cord (115 VAC models include a three-prong plug and a two-prong adapter with ground wire). Order service kits and oil below right.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Rotary vane oil vacuum pump, 4.5 cfm, 115 VAC
Max temperature 100
F (38
C),Motor type open drip-proof,Duty cycle continuous
EW-07055-04
In stock.
$765.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Economical Oil Pumps
Rotary vane oil vacuum pump, 3.75 cfm, 220 VAC
Max temperature 100
F (38
C),Motor type open drip-proof,Duty cycle continuous
EW-07055-35
In stock.
$880.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Economical Oil Pumps
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Vacuum Pumps
>
Compact/OEM Vacuum Pumps
> Economical Oil Pumps
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title