Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
GLI (11)
Product Type:
Availability:
Body material:
PVDF (12)
Glass (6)
CPVC (6)
Ryton (3)
Epoxy (3)
PEEK (1)
PPS (1)
PFA (1)
Material:
PVDF (18)
Cell type:
In-line (13)
Electrode material:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  pH Meters and Water Quality Products > Electrodes, Industrial

Electrodes, Industrial

Electrodes, Industrial
Choose from our comprehensive line of Electrodes, Industrial and other pH Meters and Water Quality Products products.
 Product Detail
Sort
 Availability
 Price
EW-27003-00 Cole-Parmer in-line/submesible flat surfaced pH electrodes

Cole-Parmer self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC

Product Type pH Electrode,ATC element None,Max temperature 176°F (80°C)
EW-27003-00
In stock.
$127.00 USD/EACH

Qty:

Representative photo only Cole Parmer self cleaning pH electrode CPVC housing 100 Pt RTD 10 ft cable tinned ends
EW-27003-12
In stock.
$161.00 USD/EACH

Qty:

Representative photo only pH Electrode flat surface 3KOhm Balco thermistor ATC 3 4 NPT threads

pH Electrode, flat surface, 3KOhm Balco thermistor ATC, 3/4" NPT threads

ATC element 3KOhm Balco thermistor,Max pressure 100 psi,Model 3-2724-10
EW-56561-02
In stock.
$303.00 USD/EACH

Qty:

EW-27400-00 Differential pH electrode

GLI Differential pH electrode, 10-ft cable, LCP body

ATC element NTC 300 thermistor,Material LCP (Liquid crystal polymer),Max pressure 100 psi
EW-27400-00
In stock.
$796.00 USD/EACH

Qty:

EW-27001-80 Cole-Parmer submersible 3-ft pH electrode

Cole-Parmer 3-ft pH electrode, submersible, double-junction, no ATC, 10-ft cable, BNC

Product Type pH Electrode,ATC element None,Brand Cole-Parmer
EW-27001-80
In stock.
$162.00 USD/EACH

Qty:

EW-27020-00 Cole-Parmer Tuff-Tip Semi-Domed pH Electrodes

Cole-Parmer Tuff-Tip Semi-Domed 3/4" In-Line or Submersible pH Electrode, No ATC, BNC

Product Type pH Electrode,ATC element None,Max temperature 230ºF(110ºC)
EW-27020-00
In stock.
$188.00 USD/EACH

Qty:

EW-27003-02 Cole-Parmer in-line/submesible flat surfaced pH electrodes

Cole-Parmer self-cleaning pH electrode, CPVC housing, 100 Pt RTD, 10-ft cable, BNC

Product Type pH Electrode,ATC element 100 Pt RTD,Max temperature 176°F (80°C)
EW-27003-02
In stock.
$161.00 USD/EACH

Qty:

EW-27301-21 Cole-Parmer in-line/submersible pH electrodes with probe guards
EW-27301-21
In stock.
$193.00 USD/EACH

Qty:

EW-05994-22 Cole-Parmer<log> General Purpose pH electrode, double-junction, 10-ft cable

Cole-Parmer General Purpose pH electrode, double-junction, 10-ft cable

Product Type pH Electrode,ATC element None,Max temperature 230°F (110°C)
EW-05994-22
In stock.
$122.00 USD/EACH

Qty:

EW-27001-90 Cole-Parmer in-line pH electrodes

Cole-Parmer pH electrode, in-line, double-junction, no ATC 10-ft cable, BNC

Product Type pH Electrode,ATC element None,Max temperature 212°F(100°C)
EW-27001-90
In stock.
$125.00 USD/EACH

Qty:

Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 1 0 4 to 20 mA output 3 4 NPT threads

+GF+ Signet Conductivity Cell with Integral Electronics, K = 1.0, 4 to 20 mA output, 3/4" NPT threads

Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-06
Usually ships in 10 days.
$804.00 USD/EACH

Qty:

EW-19901-00 Conductivity Cells

GLI 1000 Ohm electrodeless conductivity sensor, PVDF, 3/4

Product Type Conductivity Cell,Body material PVDF,Max temperature °C 200
EW-19901-00
Usually ships in 6 days or sooner.
$827.00 USD/EACH

Qty:

Representative photo only Cole Parmer In Line or Submersible Double Junction pH Electrode with Probe Guard 100 Ohm PT RTD Tinned Ends BNC
EW-27301-23
In stock.
$188.00 USD/EACH

Qty:

EW-05994-20 Cole-Parmer general purpose pH electrodes

Cole-Parmer General Purpose pH Electrode, 10-ft Cable

Product Type pH Electrode,ATC element None,Max temperature 212°F(100°C)
EW-05994-20
In stock.
$100.00 USD/EACH

Qty:

Representative photo only Cole Parmer Solution Grounded High Salt pH Electrode 100 Ohm PT RTD Tinned Ends BNC

Cole-Parmer Solution Grounded, High-Salt pH Electrode; 100 Ohm PT RTD (Tinned Ends), BNC

ATC element 100 Pt RTD,Applications High-salt applications,Brand Cole-Parmer
EW-27018-23
Usually ships in 14 days or sooner.
$271.00 USD/EACH

Qty:

365 results (showing 1 - 15 )
Per Page 
Home  >  pH Meters and Water Quality Products > Electrodes, Industrial

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title