Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Accumet (16)
Product Type:
Availability:
In Stock (141)
Built-in ATC:
No (111)
Yes (15)
Body type:
Glass (109)
Epoxy (99)
Meter style:
Waterproof:
Yes (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  pH Meters and Water Quality Products > Electrodes, Laboratory

Electrodes, Laboratory

Electrodes, Laboratory

pH Electrode Basics
The Electrode Pair
Sensing half-cell electrodes are the measuring portion of the electrode system and contain the pH sensitive membrane. Reference half-cells electrodes provide the reference potential needed for pH
measurement. Most of the electrodes available at Cole-Parmer are combination electrodes that house both half-cells in a single probe.

Bulb Material And Design
The bulb or membrane is constructed of different types of glass, each with different
useful properties.

General-purpose glass: various pH ranges, temperatures to 212ºF (100ºC).

Blue glass: pH 0 to 14, temperatures to 230ºF (110ºC).

Amber glass: pH 0 to 14, temperatures to 230ºF (110ºC), prevents Na+ error in high Na+ concentration solutions.

The bulb shape can be modified to provide a semi-dome for increased ruggedness, a spear tip for penetration applications, or even a flat surface.

Glass vs Epoxy Body
Epoxy-body electrodes are impact resistant and are ideal for rough handling but should not be used at higher temperatures or for solvents.
Note: Even though the body of an epoxy electrode is plastic, the bulb will still be glass. Glass-body electrodes withstand high temperatures and highly corrosive materials or solvents.

Sealed vs Refillable
Sealed (also known as gel-filled) electrodes are easier to maintain than refillable liquid filled electrodes. However, they must be replaced when the fill material level is low. Refillable electrodes are long-lasting and provide a faster response.

Single- vs Double-junction
In combination electrodes, the reference junction allows H+ ions to pass freely between reference and sensing half-cells to complete the electrical circuit. A singlejunction design has one place where the H+ ions pass between half-cells. The junction is located so that contaminants in your sample can also pass into the reference chamber, leaving the potential for electrode failure due to reference contamination. A double-junction design restricts the reference behind a second junction; thus, it is twice as hard for reference contamination to occur.

Selecting The Right Electrode
Cole-Parmer offers a variety of laboratory electrodes for your general-purpose and specialty laboratory applications. The “Application Guide” below provides information to help you find the electrode that best fits your application. The guide only offers suggestions—call our Application Specialists for more specific
recommendations or clarification.

Application Guide
Biological samples, proteins, and tris buffer: Double-junction or ROSS®
Pharmaceuticals: Double-junction or ROSS
Hydrofluoric acid: Antimony or HF electrode
Low ionic strength samples and acid rain: AccuFlow™ or Sure-Flow® styles
Boiler feed water and distilled water: AccuFlow or Sure-Flow styles
Drinking water: Standard Ag/AgCl with single-junction
Wastewater: Double-junction or ROSS
Solutions with heavy metals: Double-junction
Soil samples: Soil electrode or double-junction
pH>9 and high Na+: Amber glass bulb, Ag/AgCl
High or rapidly changing temperatures: ROSS styles
Moist, flat surfaces: Flat-surface
Cheese, agar, paper, and skin: Flat-surface
Semisolid samples: Spear-tip, Ag/AgCl, ISFET
Fruits, cheese, and meat: Spear-tip, Ag/AgCl, ISFET
Nonaqueous samples, solvents, and alcohols: AccuFlow styles, Sure-Flow styles, or double-junction
Viscous samples, slurries, suspended solids, and sludges: AccuFlow styles, Sure-Flow styles, double junction, or ISFET
Emulsions and oils: AccuFlow styles, Sure-Flow styles, double-junction, or ISFET
Paints and inks: AccuFlow styles, Sure-Flow styles, double-junction, or ISFET
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton pH electrode epoxy body sealed single junction general purpose 3 ft 0 9 m cable BNC connector
EW-35801-00
In stock.
$45.90 USD/EACH

Qty:

Representative photo only pH electrode combination double junction gel filled BNC connector
EW-59001-70
In stock.
$84.00 USD/EACH

Qty:

Representative photo only pH electrode combination single junction gel filled BNC connector
EW-59001-65
In stock.
$70.00 USD/EACH

Qty:

EW-35805-04 Oakton pH electrode, glass body, refillable, double-junction, flushable junction, 3-ft (0.9-m) cable, BNC connector
EW-35805-04
In stock.
$128.00 USD/EACH

Qty:

EW-55502-02 ACCUMET<REG> ACCUTUPH<REG> PH ELECTRODE, COMBINATION, DOUBLE-JUNCTION, REFILLABLE, GLASS, BNC

accumet accuTupH pH Electrode, Combination, Double-junction, Refillable, Glass, BNC

Product Type pH Electrode,Body type Glass,Brand Accumet
EW-55502-02
In stock.
$183.00 USD/EACH

Qty:

Representative photo only Cole Parmer Combination Ion Selective Electrodes Fluoride F

Cole-Parmer® Combination Ion Selective Electrodes, Fluoride (F)

Product Type pH Electrode,Brand Cole-Parmer
EW-27504-14
In stock.
$278.00 USD/EACH

Qty:

EW-27502-19 Cole-Parmer<small><sup>&reg;</sup></small> Ion-selective electrodes

Cole-Parmer ISE electrode, fluoride, double-junction, BNC

Product Type pH Electrode,Body type Glass,Built-in ATC No
EW-27502-19
In stock.
$322.00 USD/EACH

Qty:

EW-05991-36 Electrode pH/Temp double-junction for high pH, low sodium error with 3-ft cable and ATC plug for Oakton pH5/6, 11/110, 510, 700, 1100/2100, 2700 meters
In stock.
$202.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Orion fluoride electrode ionplus Sure Flow with WP BNC connector

Thermo Scientific Orion fluoride electrode ionplus Sure-Flow with WP BNC connector

Product Type Solid-State,Model 9609BNWP,Brand Ionplus
EW-05723-02
In stock.
$776.00 USD/EACH

Qty:

EW-27502-13 Cole-Parmer<small><sup>&reg;</sup></small> Ion-selective electrodes

Cole-Parmer ISE electrode, chloride, double-junction, BNC

Product Type pH Electrode,Body type Glass,Built-in ATC No
EW-27502-13
In stock.
$358.00 USD/EACH

Qty:

Representative photo only Oakton pH electrode epoxy body sealed double junction general purpose 3 ft 0 9 m cable BNC connector
EW-35805-01
In stock.
$73.44 USD/EACH

Qty:

Representative photo only Cole Parmer Combination Ion Selective Electrodes Chloride Cl

Cole-Parmer® Combination Ion Selective Electrodes, Chloride (Cl)

Product Type pH Electrode,Brand Cole-Parmer
EW-27504-08
In stock.
$227.00 USD/EACH

Qty:

EW-55500-45 Accumet<small><sup>&reg;</sup></small> MicroProbe<small><sup>&#153;</sup></small> combination electrode, flexible stem 6" L
EW-55500-45
In stock.
$315.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Orion pH ATC Triode epoxy body sealed single junction built in ATC with 8 pin miniDIN connection 5 ft 1 5 m cable BNC connector
EW-58823-30
In stock.
$248.00 USD/EACH

Qty:

EW-35805-06 Image is for illustrative purposes only. Please refer to product description
EW-35805-06
In stock.
$74.00 USD/EACH

Qty:

312 results (showing 1 - 15 )
Per Page 
Home  >  pH Meters and Water Quality Products > Electrodes, Laboratory

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title