Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Amresco (14)
Product Type:
Availability:
In Stock (121)
CE Compliance:
Yes (40)
No. of wells:
10 (25)
14 (20)
12 (13)
20 (10)
27 (6)
15 (3)
8/10 (3)
12/16 (3)
10/14 (3)
8/16 (3)
13 (3)
16 (3)
26 (3)
28 (3)
51 (3)
102 (3)
30 (3)
Wavelength:
302 nm (22)
254 nm (11)
Buffer solution required (mL):
1600 (16)
250 (3)
1000 (2)
2300 (2)
700 (2)
3000 (2)
400 (1)
100 (1)
4500 (1)
1400 (1)
410 (1)
450 (1)
800 (1)
150 (1)
750 (1)
1500 (1)
725 (1)
500 (1)
825 (1)
900 (1)
Gel thickness (mm):
1.5 (16)
0.8 (2)
0.4 (2)
Gel size (cm L):
8 (10)
25 (5)
10 (4)
45 (2)
14 (1)
40 (1)
8.5 (1)
16 (1)
20 (1)
26 (1)
Max current:
500 mA (9)
400 mA (5)
300 mA (4)
Gel size (cm W):
8.5 (9)
20 (3)
14 (3)
10 (2)
13 (2)
23 (2)
7 (1)
12 (1)
35 (1)
5.5 (1)
7.5 (1)
26 (1)
Qty/pk:
100 (6)
Max voltage:
200 V (3)
1000V (3)
3000V (3)
300V (2)
250VDC (2)
150 (1)
Ash:
0.06% (4)
0.01% (2)
Size:
100g (3)
25g (2)
500g (1)
250g (1)
25 g (1)
100 g (1)
250 g (1)
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Electrophoresis Products

Electrophoresis Products

Cole-Parmer offers a complete line of products for electrophoresis including horizontal and vertical gel electrophoresis systems, reagents, imaging systems, and power supplies. Along with our wide range of products, we also offer expert assistance in finding the right product. For more information or help choosing the right product for your need, contact our Application Specialists.

Gel electrophoresis is a method by which DNA, RNA or proteins are separated by electric current typically in an agarose gel. Agarose gels are cast and placed in a gel box with buffer. Sample is pipetted into wells cast into the gels and the electric current is applied to the system. The electric current causes the molecules to move through the gel at different rates depending on their mass and mass to charge ratio. Once complete, staining of the molecules within the gel allow for visibility by the researcher. If the molecules are able to fluoresce, an image of the gel and molecular movement can be taken.

Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
21700_00.jpg - Cole-Parmer<reg> Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk

Cole-Parmer Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk

Product Type Electrophoresis Accessories,Brand Cole-Parmer
EW-21700-03
In stock.
$105.00 USD/PACKAGE

Qty:

28403_60.jpg - Programmable Power Supply, 600 VDC/1000 mA, 230V

Programmable Power Supply, 600 VDC/1000 mA, 230V

Max current 1000 mA,Max voltage 600 VDC,Dimensions " W 12-1/4
EW-28403-86
Usually ships in 35 days.
$2,060.00 USD/EACH

Qty:

28406SE.jpg - Thermo Scientific Electrophoresis Power Supply 3000v 120v

Thermo Scientific Electrophoresis Power Supply 3000v 120v

Max current 400 mA,Max voltage 3000V,Dimensions " W 5-½"W
EW-28406-50
Usually ships in 30 days.
$2,475.75 USD/EACH

Qty:

PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It Imaging System with 302nm 20cm x 20cm Filter Transilluminator, 115VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-14
Usually ships in 11 days.
$3,497.00 USD/EACH

Qty:

PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It Imaging System with 302nm 20cm x 20cm Filter Transilluminator, 230VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-16
Usually ships in 28 days.
$3,497.00 USD/EACH

Qty:

17314_11.jpg - Enduro Gel XL Electrophoresis System 120V
EW-17314-11
Usually ships in 5 days.
$580.00 USD/EACH

Qty:

0664580.jpg - Nitrocellulose membranes 0.45um 300cm/roll

Nitrocellulose membranes 0.45um 300cm/roll

Product Type Nitrocellulose Membranes,Filter size 0.45 um,Pore size micron 0.45
EW-06645-80
In stock.
$230.00 USD/ROLL

Qty:

no_image.gif - X-RAY FILM BLUE 5X7" 100/PK
EW-21700-01
In stock.
$70.00 USD/PACKAGE

Qty:

no_image.gif - Doc-It<reg> LS Imaging Software- order in addition to BioDoc-It<tm> Imaging Systems

Doc-It LS Imaging Software- order in addition to BioDoc-It Imaging Systems

Product Type Electrophoresis Imaging Systems Accessories,Brand UVP
EW-97701-62
Usually ships in 24 days.
$1,620.00 USD/EACH

Qty:

97623_08_R.jpg - Cole-Parmer 8-Watt UV Transilluminator 302nm Wavelength with 15cm x 15cm Filter, 115VAC

Cole-Parmer 8-Watt UV Transilluminator 302nm Wavelength with 15cm x 15cm Filter, 115VAC

Product Type Ultraviolet Transilluminators,Watts 8,Wavelength 302 nm
EW-97623-04
Usually ships in 20 days.
$1,100.00 USD/EACH

Qty:

97622_00_R.jpg - Cole-Parmer 25-watt Dual-Intensity 302nm Wavelength UV Transilluminator with 20cm x 20cm Filter, 230VAC

Cole-Parmer 25-watt Dual-Intensity 302nm Wavelength UV Transilluminator with 20cm x 20cm Filter, 230VAC

Product Type Ultraviolet Transilluminators,Watts 25,Wavelength 302 nm
EW-97622-06
Usually ships in 20 days.
$1,620.00 USD/EACH

Qty:

2857010.jpg - Cole-Parmer<reg> Vertical Dual Adjustable Slab Gel System

Cole-Parmer Vertical Dual Adjustable Slab Gel System

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 825,No. of wells 10, 12, 14, 20, 29, 30
EW-28570-10
Usually ships in 27 days.
$678.00 USD/EACH

Qty:

2855600.jpg - Nucleic Acid Sequencing System sequencing system20cm x 45cm

Nucleic Acid Sequencing System sequencing system20cm x 45cm

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 450,No. of wells 20
EW-28556-00
Usually ships in 14 days.
$1,045.00 USD/EACH

Qty:

PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It 50 Imaging System, 230VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-12
Usually ships in 21 days.
$2,182.00 USD/EACH

Qty:

3475075.jpg - UV PCR chamber, 110 VAC, 60 Hz

UV PCR chamber, 110 VAC, 60 Hz

Interior dimensions " W 24,Product Type PCR Enclosures,Interior dimensions " H 28
EW-34750-75
Usually ships in 21 days.
$3,070.00 USD/EACH

Qty:
See all UV PCR Chamber

400 results (showing 1 - 15 )
Per Page 

View products by: Job Title | Category | Brand | Part Number | Price
View product categories by: Popularity | Favorites | Job Title | Price | Part Number
View popular items by: Popularity | Our Choices | All-Around Favorites | Job Title
View reviews by: Popularity | Product Title | Best Rated