Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Product Type:
Availability:
No. of wells:
10 (25)
14 (20)
12 (10)
20 (8)
27 (6)
16 (5)
15 (3)
8/10 (3)
12/16 (3)
10/14 (3)
8/16 (3)
13 (3)
26 (3)
28 (3)
51 (3)
102 (3)
4 (2)
CE Compliance:
Yes (22)
Buffer solution required (mL):
1600 (16)
250 (3)
300 (3)
2300 (2)
700 (2)
400 (1)
1000 (1)
4500 (1)
1400 (1)
3000 (1)
150 (1)
750 (1)
1500 (1)
725 (1)
500 (1)
825 (1)
900 (1)
50 mL (1)
Gel thickness (mm):
1.5 (15)
Gel size (cm L):
8 (10)
25 (4)
10 (3)
40 (1)
8.5 (1)
16 (1)
20 (1)
26 (1)
Gel size (cm W):
8.5 (9)
14 (3)
13 (2)
20 (2)
7.5 (2)
7 (1)
23 (1)
5.5 (1)
26 (1)
Glass size (cm H):
10 (9)
8 (9)
14.5 (7)
16 (7)
19 (7)
22 (7)
28 (7)
Glass size (cm W):
16.5 (9)
20 (5)
16 (3)
Glass size (cm L):
20 (5)
14 (3)
14.5 (1)
16 (1)
19 (1)
22 (1)
28 (1)
Learn more:
[-] Show Less
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Life Science > Genomics > Electrophoresis Products

Electrophoresis Products

Electrophoresis Products
Choose from our comprehensive line of Electrophoresis Products and other Genomics products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Instant Blue Stain 1 liter

Instant Blue Stain, 1 liter

Product Type Electrophoresis Accessories
EW-28570-59
In stock.
$209.00 USD/EACH

Qty:

Representative photo only Programmable Power Supply 6000 VDC 150 mA 115V

Programmable Power Supply, 6000 VDC/150 mA, 115V

Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-88
Call 1-888-358-4705 for availability.
$3,490.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Electrophoresis Power Supply 3000v 120v

Thermo Scientific Electrophoresis Power Supply 3000v 120v

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-50
Call 1-888-358-4705 for availability.
$2,574.00 USD/EACH

Qty:

EW-28575-00 Cole-Parmer<small><sup>&reg;</sup></small> Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Cole-Parmer Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 250,No. of wells 10, 14
EW-28575-00
In stock.
$375.00 USD/EACH

Qty:

EW-28560-00 Midi-horizontal gel units, 14 x 10 cm gel bed

Midi-horizontal gel units, 14 x 10 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 700,No. of wells 10, 13, 16, 20, 27
EW-28560-00
Call 1-888-358-4705 for availability.
$385.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Electrophoresis Power Supply 1000v 230v UK plug

Thermo Scientific Electrophoresis Power Supply 1000v 230v UK plug

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-35
Call 1-888-358-4705 for availability.
$1,532.00 USD/EACH

Qty:

EW-28406-20 Thermo Scientific Electrophoresis Power Supply 300v 120v

Thermo Scientific Electrophoresis Power Supply 300v 120v

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-20
In stock.
$547.00 USD/EACH

Qty:

EW-28570-00 Cole-Parmer<small><sup>&reg;</sup></small> Vertical Dual Adjustable Slab Gel System

Cole-Parmer Vertical Single Adjustable Slab Gel System

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 725,No. of wells 10, 12, 14, 20, 29, 30
EW-28570-00
Call 1-888-358-4705 for availability.
$500.00 USD/EACH

Qty:

EW-28562-00 Cole-Parmer<small><sup>&reg;</sup></small> High-Throughput Horizontal Gel System

Cole-Parmer High-Throughput Horizontal Gel System

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 3000,No. of wells 26, 27, 28, 51, 102
EW-28562-00
Call 1-888-358-4705 for availability.
$522.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Electrophoresis Power Supply 300v 230v European plug

Thermo Scientific Electrophoresis Power Supply 300v 230v European plug

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-25
Usually ships in 8 days.
$619.00 USD/EACH

Qty:

EW-17314-11 Enduro Gel XL Electrophoresis System 120V

Enduro Gel XL Electrophoresis System 120V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-11
Usually ships in 5 days.
$629.50 USD/EACH

Qty:

Representative photo only Enduro Gel XL Electrophoresis System with UK plug 230V

Enduro Gel XL Electrophoresis System with UK plug 230V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-13
Usually ships in 5 days.
$629.50 USD/EACH

Qty:

Representative photo only Enduro Gel XL Electrophoresis System with EURO Plug 230V

Enduro Gel XL Electrophoresis System with EURO Plug 230V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-15
Usually ships in 5 days.
$629.50 USD/PACKAGE

Qty:

EW-28559-00 Mini-horizontal gel unit, 5.5 x 8.5 cm gel bed

Horizontal Mini-gel unit, 5.5 x 8.5 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 150,No. of wells 8/10, 12/16
EW-28559-00
Call 1-888-358-4705 for availability.
$255.00 USD/EACH

Qty:

Representative photo only Cole Parmer 4 place Electrophoresis System with casting kit 14 well 0 75 mm comb thickness

Cole-Parmer 4-place Electrophoresis System with casting kit, 14 well, 0.75 mm comb thickness

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 1600,No. of wells 14
EW-28571-28
Usually ships in 6 days.
$720.00 USD/EACH

Qty:

286 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Genomics > Electrophoresis Products

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title