Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Life Science > Electrophoresis Products > Electrophoresis Systems - Horizontal Configuration

Electrophoresis Systems - Horizontal Configuration

Electrophoresis Systems - Horizontal Configuration
Horizontal gel electrophoresis systems and other electrophoresis products are now available from the Cole-Parmer catalog online. Click any horizontal gel electrophoresis system for more information.
 Product Detail
Sort
 Availability
 Price
EW-28559-00 Mini-horizontal gel unit, 5.5 x 8.5 cm gel bed

Horizontal Mini-gel unit, 5.5 x 8.5 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 150,No. of wells 8/10, 12/16
EW-28559-00
Call 1-888-358-4705 for availability.
$255.00 USD/EACH

Qty:

EW-28560-00 Midi-horizontal gel units, 14 x 10 cm gel bed

Midi-horizontal gel units, 14 x 10 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 700,No. of wells 10, 13, 16, 20, 27
EW-28560-00
Call 1-888-358-4705 for availability.
$385.00 USD/EACH

Qty:

EW-28562-00 Cole-Parmer<small><sup>&reg;</sup></small> High-Throughput Horizontal Gel System

Cole-Parmer High-Throughput Horizontal Gel System

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 3000,No. of wells 26, 27, 28, 51, 102
EW-28562-00
Call 1-888-358-4705 for availability.
$522.00 USD/EACH

Qty:

EW-17314-11 Enduro Gel XL Electrophoresis System 120V

Enduro Gel XL Electrophoresis System 120V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-11
Usually ships in 5 days.
$629.50 USD/EACH

Qty:

Representative photo only Enduro Gel XL Electrophoresis System with UK plug 230V

Enduro Gel XL Electrophoresis System with UK plug 230V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-13
Usually ships in 5 days.
$629.50 USD/EACH

Qty:

Representative photo only Enduro Gel XL Electrophoresis System with EURO Plug 230V

Enduro Gel XL Electrophoresis System with EURO Plug 230V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-15
Usually ships in 5 days.
$629.50 USD/PACKAGE

Qty:

EW-28559-10 Mini-horizontal gel unit, 7.5 x 10 cm gel bed

Horizontal Mini-gel unit, 7.5 x 10 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 250,No. of wells 10/14, 8/16
EW-28559-10
Call 1-888-358-4705 for availability.
$300.00 USD/EACH

Qty:

Representative photo only Midi horizontal gel units 14 x 16 cm gel bed

Midi-horizontal gel units, 14 x 16 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 750,No. of wells 10, 13, 16, 20, 27
EW-28560-10
In stock.
$390.00 USD/EACH

Qty:

Representative photo only Non recirculating cell 13 x 25 cm gel size

Non-recirculating cell, 13 x 25 cm gel size

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 1600,No. of wells 12, 16, 20
EW-28553-00
Usually ships in 9 days.
$682.00 USD/EACH

Qty:

Representative photo only Recirculating Cell 23 x 40 cm gel size

Recirculating Cell, 23 x 40 cm gel size

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 4500,No. of wells 25, 50
EW-28553-40
Usually ships in 14 days.
$846.00 USD/EACH

Qty:

Representative photo only Recirculating Cell 20 x 25 cm gel size

Recirculating Cell, 20 x 25 cm gel size

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 2300,No. of wells 16, 24, 36
EW-28553-30
Usually ships in 4 days.
$818.00 USD/EACH

Qty:

Representative photo only Non recirculating cell 20 x 25 cm gel size

Non-recirculating cell, 20 x 25 cm gel size

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 2300,No. of wells 16, 24, 36
EW-28553-20
Usually ships in 6 days.
$736.00 USD/EACH

Qty:

EW-28553-10 Recirculating Cell, 13 x 25 cm gel size

Recirculating Cell, 13 x 25 cm gel size

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 1600,No. of wells 12, 16, 20
EW-28553-10
Usually ships in 4 days.
$740.00 USD/EACH

Qty:

EW-28550-00 Mini horizontal cell

Mini horizontal cell

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 400,No. of wells 6, 10
EW-28550-00
Usually ships in 6 days.
$388.00 USD/EACH

Qty:

EW-28561-00 Cole-Parmer<small><sup>&reg;</sup></small> Submarine Horizontal Gel System

Cole-Parmer Submarine Horizontal Gel System

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 1500,No. of wells 10, 13, 16, 20, 27
EW-28561-00
Call 1-888-358-4705 for availability.
$495.00 USD/EACH

Qty:

15 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Electrophoresis Products > Electrophoresis Systems - Horizontal Configuration

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title