Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Product Type:
Availability:
No. of wells:
10 (12)
14 (11)
12 (3)
20 (3)
15 (3)
30 (2)
29 (2)
10, 14 (2)
24 (1)
15, 20 (1)
Gel thickness (mm):
1.5 (9)
Glass size (cm H):
10 (9)
8 (9)
14.5 (7)
16 (7)
19 (7)
22 (7)
28 (7)
Glass size (cm W):
16.5 (9)
20 (5)
16 (3)
Glass size (cm L):
20 (5)
14 (3)
14.5 (1)
16 (1)
19 (1)
22 (1)
28 (1)
Buffer solution required (mL):
250 (2)
700 (1)
1400 (1)
725 (1)
500 (1)
825 (1)
900 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Life Science > Electrophoresis Products > Electrophoresis Systems - Vertical Configuration

Electrophoresis Systems - Vertical Configuration

Electrophoresis Systems - Vertical Configuration
Vertical electrophoresis systems and other gel electrophoresis products are now available to order from the Cole-Parmer online catalog. Click any vertical gel electrophoresis system for more information.
 Product Detail
Sort
 Availability
 Price
EW-28575-00 Cole-Parmer<small><sup>&reg;</sup></small> Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Cole-Parmer Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 250,No. of wells 10, 14
EW-28575-00
In stock.
$375.00 USD/EACH

Qty:

EW-28570-00 Cole-Parmer<small><sup>&reg;</sup></small> Vertical Dual Adjustable Slab Gel System

Cole-Parmer Vertical Single Adjustable Slab Gel System

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 725,No. of wells 10, 12, 14, 20, 29, 30
EW-28570-00
Call 1-888-358-4705 for availability.
$500.00 USD/EACH

Qty:

EW-28570-15 Cole-Parmer<small><sup>&reg;</sup></small> Vertical Dual Adjustable Slab Gel System

Cole-Parmer Vertical Dual Adjustable Slab Gel System, CE-marked

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 900,No. of wells 10, 12, 14, 20, 29, 30
EW-28570-15
Call 1-888-358-4705 for availability.
$620.00 USD/EACH

Qty:

Representative photo only Comb for Cole Parmer Dual Vertical Mini Gel System Model 28575 00 10 Wells 0 5 mm Thick

Comb for Cole-Parmer Dual Vertical Mini-Gel System Model 28575-00; 10 Wells, 0.5 mm Thick

Product Type Electrophoresis Combs,No. of wells 10,Brand Cole-Parmer
EW-28575-50
In stock.
$30.00 USD/EACH

Qty:

Representative photo only Glass Plate Set for Cole Parmer Vertical Slab Gel Systems 22 cm Plate Height

Glass Plate Set for Cole-Parmer Vertical Slab Gel Systems, 22 cm Plate Height

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm W 16.5,Glass size cm H 22
EW-28573-63
Call 1-888-358-4705 for availability.
$42.00 USD/EACH

Qty:

Representative photo only Spacer Set for Cole Parmer Dual Vertical Mini Gel System Model 28575 00 0 5 mm

Spacer Set for Cole-Parmer Dual Vertical Mini-Gel System Model 28575-00, 0.5 mm

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm H 10,Brand Cole-Parmer
EW-28575-70
Call 1-888-358-4705 for availability.
$18.50 USD/EACH

Qty:

Representative photo only Comb for Cole Parmer Vertical Slab Gel System 30 Wells 1 0 mm Thick

Comb for Cole-Parmer Vertical Slab Gel System; 30 Wells, 1.0 mm Thick

Product Type Electrophoresis Combs,No. of wells 30,Brand Cole-Parmer
EW-28572-61
In stock.
$45.00 USD/EACH

Qty:

Representative photo only Glass Plate Set for Cole Parmer Vertical Slab Gel Systems 28 cm Plate Height

Glass Plate Set for Cole-Parmer Vertical Slab Gel Systems, 28 cm Plate Height

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm W 16.5,Glass size cm H 28
EW-28573-64
In stock.
$44.00 USD/EACH

Qty:

Representative photo only Glass Plate Set for Cole Parmer Dual Vertical Mini Gel System Model 28575 00

Glass Plate Set for Cole-Parmer Dual Vertical Mini-Gel System Model 28575-00

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm W 11.3,Glass size cm H 10
EW-28575-90
In stock.
$23.00 USD/EACH

Qty:

Representative photo only Spacer Set for Cole Parmer Vertical Slab Gel Systems 14 5 cm Height 1 0 mm Thick

Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 14.5 cm Height, 1.0 mm Thick

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm H 14.5,Brand Cole-Parmer
EW-28573-05
In stock.
$30.00 USD/EACH

Qty:

Representative photo only Spacer Set for Cole Parmer Dual Vertical Mini Gel System Model 28575 00 0 75 mm

Spacer Set for Cole-Parmer Dual Vertical Mini-Gel System Model 28575-00, 0.75 mm

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm H 10,Brand Cole-Parmer
EW-28575-71
In stock.
$18.50 USD/EACH

Qty:

Representative photo only Spacer Set for Cole Parmer Vertical Slab Gel Systems 22 cm Height 1 0 mm Thick

Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 22 cm Height, 1.0 mm Thick

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm H 22,Brand Cole-Parmer
EW-28573-08
Call 1-888-358-4705 for availability.
$31.00 USD/EACH

Qty:

Representative photo only Spacer Set for Cole Parmer Vertical Slab Gel Systems 28 cm Height 1 5 mm Thick

Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 28 cm Height, 1.5 mm Thick

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm H 28,Brand Cole-Parmer
EW-28573-14
In stock.
$32.00 USD/EACH

Qty:

Representative photo only Comb for Cole Parmer Vertical Slab Gel System 10 Wells 1 0 mm Thick

Comb for Cole-Parmer Vertical Slab Gel System; 10 Wells, 1.0 mm Thick

Product Type Electrophoresis Combs,No. of wells 10,Brand Cole-Parmer
EW-28572-56
In stock.
$40.00 USD/EACH

Qty:

Representative photo only Spacer Set for Cole Parmer Vertical Slab Gel Systems 14 5 cm Height 0 75 mm Thick

Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 14.5 cm Height, 0.75 mm Thick

Product Type Electrophoresis Vertical Systems Accessories,Glass size cm H 14.5,Brand Cole-Parmer
EW-28573-00
In stock.
$30.00 USD/EACH

Qty:

106 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Electrophoresis Products > Electrophoresis Systems - Vertical Configuration

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title