log in or register
Price Range:
Availability:
Max volume (mL):
5 (1)
10 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fluid & Liquid Handling > Pipettes and Pipettors > Single-Channel Pipettors > Eppendorf Pipettors > Eppendorf Research plus Adjustable-Pipettors
Eppendorf Research plus Adjustable-Pipettors
Eppendorf Research plus Adjustable-Pipettors
  • Requires up to 50% less pipetting force, designed to minimize stress and fatigue
  • The ultralight pipette is designed to fit comfortably in your hand
  • Made with industry-leading materials for exceptional durability
The Research plus was optimized for form and function. The ultralightweight pipette meets the highest needs for precision and accuracy- combined with ultimate ergonomics and increased flexibility. The spring-loaded tip cone achieves optimal tightness and minimal attachment forces - doing most of the work for you! Make your volume adjustments with just one hand, only a few turns will get you to your desired volume. The four-digit volume display is easy to view. The multichannel pipettors offer channel indicators, in addition to flexibility of removing any of the independent channels - use as many as you need.
Disassemble your pipettor with ease - the quick connection clip lets you remove the lower part in just two steps. The secondary adjustment gives you greater accuracy for specific liquid types. Pipettors are made from Fortron® PPS, which is resistant to acids, alkalis, mildew, bleaches, aging, sunlight, heat, and abrasion. Fully autoclavable for simple decontamination.
 Product Detail
Sort
 Availability
 Price
Representative photo only Single channel Eppendorf Research plus manual pipette 100 1000 L blue control button
EW-24505-08
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 1 10 mL turquoise control button
EW-24505-11
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 0 5 5 mL violet control button
EW-24505-09
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 10 100 L yellow control button
EW-24505-05
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 20 200 L yellow control button
EW-24505-06
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 0 5 10 L gray control button
EW-24505-02
Usually ships in 10 days or sooner.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 2 20 L yellow control button
EW-24505-04
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Twelve channel Eppendorf Research plus manual pipette 10 100 L yellow control button
EW-24505-16
Usually ships in 13 days.
$974.00 USD/EACH
Qty:
Representative photo only Eight channel Eppendorf Research plus manual pipette 10 100 L yellow control button
EW-24505-13
Usually ships in 13 days.
$841.00 USD/EACH
Qty:
Representative photo only Twelve channel Eppendorf Research plus manual pipette 30 300 L orange control button
EW-24505-17
Usually ships in 13 days.
$974.00 USD/EACH
Qty:
Representative photo only Twelve channel Eppendorf Research plus manual pipette 0 5 10 L gray control button
EW-24505-15
Usually ships in 13 days.
$974.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 2 20 L light gray control button
EW-24505-03
Usually ships in 13 days.
$348.00 USD/EACH
Qty:
Representative photo only Eight channel Eppendorf Research plus manual pipette 30 300 L orange control button
EW-24505-14
Usually ships in 13 days.
$841.00 USD/EACH
Qty:
EW-24505-01 Single-channel Eppendorf Research<small><sup>&reg;</sup></small> plus manual pipette 0.1-2.5 &micro;L, dark gray control button
EW-24505-01
In stock.
$348.00 USD/EACH
Qty:
Representative photo only Single channel Eppendorf Research plus manual pipette 30 300 L orange control button
EW-24505-07
Usually ships in 13 days.
$348.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
Home  >  Fluid & Liquid Handling > Pipettes and Pipettors > Single-Channel Pipettors > Eppendorf Pipettors > Eppendorf Research plus Adjustable-Pipettors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title