Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Brand:
Price Range:
Availability:
Quantity (mL):
500 (2)
4000 (2)
1000 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Ethyl Alcohol Denatured

Ethyl Alcohol Denatured
Choose from our comprehensive line of Ethyl Alcohol Denatured and other Ethaned to Ethyla products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Ethanol Anhydrous Denatured 200 Proof 4L

Ethanol, Anhydrous Denatured 200 Proof 4L

Product Type Chemical Reagents,Form Liquid,Hazard symbol Flammable
EW-89000-59
Usually ships in 5 days.
$85.00 USD/EACH

Qty:

Representative photo only Ethanol Anhydrous Denatured 200 Proof 500mL

Ethanol, Anhydrous Denatured 200 Proof 500mL

Product Type Chemical Reagents,Form Liquid,Hazard symbol Flammable
EW-89000-57
Usually ships in 6 days.
$25.00 USD/EACH

Qty:

Representative photo only Ethyl Alcohol Denatured Proprietary Solvent 4 liter

Ethyl Alcohol Denatured (Proprietary Solvent) (4 liter)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Flammable
EW-88008-66
In stock.
$155.00 USD/EACH

Qty:

Representative photo only Ethyl Alcohol Denatured Proprietary Solvent 1 liter

Ethyl Alcohol Denatured (Proprietary Solvent) (1 liter)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Flammable
EW-88008-64
In stock.
$64.00 USD/EACH

Qty:

Representative photo only Ethyl Alcohol Denatured Proprietary Solvent 500mL

Ethyl Alcohol Denatured (Proprietary Solvent) (500mL)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Flammable
EW-88008-62
In stock.
$45.00 USD/EACH

Qty:

5 results (showing 1 - 5 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title