Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Quantity (mL):
4000 (4)
20000 (1)
200000 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Ethyl alcohol

Ethyl alcohol
Choose from our comprehensive line of Ethyl alcohol and other Ethaned to Ethyla products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Ethyl Alcohol Denatured 20l

Ethyl Alcohol Denatured 20l

Product Type Chemical Reagents,Form Liquid,Quantity mL 20000
EW-88051-70
In stock.
$370.00 USD/EACH

Qty:
See all Ethyl alcohol

Representative photo only Ethanol Cda19 Histo F sty 1gal

Ethanol Cda19 Histo F-sty 1gal

Product Type Chemical Reagents,Form Liquid
EW-88051-68
In stock.
$170.00 USD/EACH

Qty:
See all Ethyl alcohol

Representative photo only Ethyl Alcohol Denatured 4l

Ethyl Alcohol Denatured 4l

Product Type Chemical Reagents,Form Liquid,Quantity mL 4000
EW-88051-74
In stock.
$153.00 USD/EACH

Qty:
See all Ethyl alcohol

Representative photo only Ethyl alcohol denatured for HPLC 4l

Ethyl alcohol, denatured for HPLC (4l)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Highly_Flammable
EW-88364-81
In stock.
$200.00 USD/EACH

Qty:
See all Ethyl alcohol

Representative photo only Ethyl Alc Denatur 4l Safe tin

Ethyl Alc Denatur 4l Safe-tin

Product Type Chemical Reagents,Form Liquid,Quantity mL 4000
EW-88051-76
In stock.
$183.00 USD/EACH

Qty:
See all Ethyl alcohol

Representative photo only Ethyl Alc Denatur 4l Safecote

Ethyl Alc Denatur 4l Safecote

Product Type Chemical Reagents,Form Liquid,Quantity mL 4000
EW-88051-77
In stock.
$169.00 USD/EACH

Qty:
See all Ethyl alcohol

Representative photo only Ethyl Alcohol 200l

Ethyl Alcohol 200l

Product Type Chemical Reagents,Form Liquid,Quantity mL 200000
EW-88051-71
In stock.
$2,061.00 USD/EACH

Qty:
See all Ethyl alcohol

7 results (showing 1 - 7 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title