United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
Conductivity/Resistivity/TDS Control
>
1/8-DIN Conductivity Control
> Eutech Instruments COND 200 1/8-DIN Economical Conductivity Controller
Eutech Instruments COND 200 1/8-DIN Economical Conductivity Controller
Economical controller designed with the features and reliability of a much more expensive instrument
Intuitive user interface; scrolling 14-segment LED display makes setup simple
Reliable power supply withstands voltage fluctuation from 85 to 265 VAC
Adjustable hysteresis prevents rapid relay switching near set points
This sturdy, compact controller features two relays for low/high, low/low or high/high controlactivate an alarm, pump or other device. The controller features 4 to 20 mA transmitter/recorder output for datalogging, and adjustable temperature coefficient and selectable cell constants for best accuracy in a wide variety of solutions. Range selection from menu allows the user to define range without the use of dip switches.
Two-level password protection allows user to access all controller functions or just calibration features, which prevents tampering with setup functions. Built-in memory backup retains setup even if power fails, and lets the user configure unit before installation. Removable terminal strips make connections quick and easy. Use with 0.1 and 1.0 cell constan
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Eutech Instruments 1/8-DIN COND 200, On/Off Controller
Control type On/Off,Display 4-digit, 14-segment LED
EW-19505-00
In stock.
$449.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Eutech Instruments COND 200 1/8-DIN Economical Conductivity Controller
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
Conductivity/Resistivity/TDS Control
>
1/8-DIN Conductivity Control
> Eutech Instruments COND 200 1/8-DIN Economical Conductivity Controller
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title