Home  >  pH Meters and Water Quality Products > Process pH and Water Quality Control > Conductivity/Resistivity/TDS Control > 1/4-DIN Conductivity Control > Eutech Instruments COND 500 Industrial Conductivity Transmitter with Display
Eutech Instruments COND 500 Industrial Conductivity Transmitter with Display
Eutech Instruments COND 500 Industrial Conductivity Transmitter with Display
  • Economical and compact—yet ideal for tough applications
  • Weatherproof IP65-rated housing—suitable for harsh environments
  • Displays conductivity and temperature
  • Two wire design with input-to-output isolation
Simple and reliable transmitters are ideal for connecting to PLC for advanced control systems. Or, connect to a recorder for a low cost documentation system. Scaleable 4-20 mA output with automatic temperature compensation.
Install in series with each transmitter isolated so there is no disruption if one should fail. Housing contains mounting holes for easy installation on plates, poles, or walls. Knockouts with rubber septums allow wires to be connected from any direction, but keep dust and moisture out. Order conductivity cells separately from accessories.
 Product Detail
Sort
 Availability
 Price
Representative photo only Eutech Instruments 1 4 DIN COND 500 Industrial Conductivity Transmitter with Display
EW-19505-20
In stock.
$436.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  pH Meters and Water Quality Products > Process pH and Water Quality Control > Conductivity/Resistivity/TDS Control > 1/4-DIN Conductivity Control > Eutech Instruments COND 500 Industrial Conductivity Transmitter with Display
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title