United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
CE Compliance:
Yes
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
1/4-DIN pH/ORP Control
> Eutech Instruments pH 1000 1/4-DIN pH/ORP Controller/Transmitter
Eutech Instruments pH 1000 1/4-DIN pH/ORP Controller/Transmitter
All the features of the economical pH 800 plus proportional control
Controller features fully adjustable 0 to 20 mA/4 to 20 mA output
Easily program relays, either to normally open or normally closed, with the front keypad
Two level password protection keeps your program parameters safe
Control your process with three available control strategies: On/off, proportional pulse length, or proportional pulse frequency. Use proportional control to gradually increase or decrease pump or valve response, depending on the deviation from the set point.
Manually control test relays with the touch of one button. Relays include programmable time delay to minimize false alarms. Hold relay feature lets you connect other controllers, flow switches, or level switches for failsafe operation.
Controller is compatible with a variety of electrodes, including glass and antimony. Symmetrical electrode inputs eliminate electrical noise problems when used with a solution ground. Connections are made through the two removable terminal strips for easy installation. Order electrodes separately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Eutech Instruments pH 1000 controller, 115 VAC
Product Type Controller,Accuracy pH -0.01 to 0.01,Resolution pH 0.01
EW-56710-00
In stock.
$730.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Eutech Instruments pH 1000 1/4-DIN pH/ORP Controller/Transmitter
Eutech Instruments pH 1000 controller, 220 VAC
Product Type Controller,Accuracy pH -0.01 to 0.01,Resolution pH 0.01
EW-56710-05
Call 1-888-358-4705 for availability.
$730.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Eutech Instruments pH 1000 1/4-DIN pH/ORP Controller/Transmitter
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
1/4-DIN pH/ORP Control
> Eutech Instruments pH 1000 1/4-DIN pH/ORP Controller/Transmitter
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title