United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
1/8-DIN pH/ORP Control
> Eutech Instruments pH 190 1/8-DIN pH/ORP Controller/Transmitter
Eutech Instruments pH 190 1/8-DIN pH/ORP Controller/Transmitter
Improved user interface allows direct access to calibration mode and set point adjustment
Universal power supply input from 85 to 250 VAC/DC
Automatic temperature compensation
Galvanically isolated 4 to 20 mA output
The pH 190 1/8-DIN controller displays the measured parameter value or temperature with a touch of a button. The user interface of the popular pH 200 series was simplified for quick set point review and programming. Entrance into the calibration mode is accomplished by simply pressing keys on the front panel--no need to remember and enter security codes. All information is displayed on the 14 segment LED display that is easy to read in low light installations.
The controller has two relays that can be set for any combination of low/high level on/off (limit) control. Independently adjustable hysteresis provide accurate control in varying applications and prevents relay chatter near the set-point. All data is retained in non-volatile memory.
The controller can operate in asymmetrical or symmetrical mode operation. Asymmetrical mode uses standard industrial pH electrodes and is acceptable for environments that are not electrically noisy. Symmetrical mode eliminates electronic noise problems when using electrodes with solution grounds. Order 100
Pt RTD pH or ORP electrodes with BNC pH connections seperately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Eutech Instruments pH 190 1/8-DIN pH/ORP controller
Product Type Controller,Accuracy pH -0.01 to 0.01,Resolution pH 0.01
EW-56700-10
In stock.
$396.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Eutech Instruments pH 190 1/8-DIN pH/ORP Controller/Transmitter
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
1/8-DIN pH/ORP Control
> Eutech Instruments pH 190 1/8-DIN pH/ORP Controller/Transmitter
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title