log in or register
Product Type:
Price Range:
Availability:
hp:
1/2 (3)
3/4 (3)
1/4 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Motors > AC Single and Three Motors for Positive Displacement Pumps > Explosion-Proof NEMA Type C-Face Motors
Explosion-Proof NEMA Type C-Face Motors
Explosion-Proof NEMA Type C-Face Motors
  • For Class 1 Group C and D, Class 2 Group F and G locations
  • Automatic overload protection stops and restarts motor in case of overload or overheating
  • UL-listed file no. E-12044
Motors are TEFC (models 70007-00 to -12 are TENV) and include an explosion-proof conduit box (shipped loose).
 Product Detail
Sort
 Availability
 Price
Representative photo only Explosion proof Single phase NEMA Type C face Motor 1 Hp 1800 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 1 Hp, 1800 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 1,Model G870
EW-71006-45
Usually ships in 15 days.
$704.00 USD/EACH
Qty:
Representative photo only Explosion proof Three phase NEMA Type C face Motor 1 2 Hp 1800 RPM 208 230 460 VAC
Explosion-proof Three-phase NEMA Type C-face Motor, 1/2 Hp, 1800 RPM, 208-230/460 VAC
Product Type Three-phase explosion-proof motors,hp 1/2,Model 56T17G5315/ G829
EW-71007-25
Usually ships in 6 days.
$604.00 USD/EACH
Qty:
Representative photo only Explosion proof Single phase NEMA Type C face Motor 1 2 Hp 1800 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 1/2 Hp, 1800 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 1/2,Model G855
EW-71006-25
Usually ships in 14 days.
$604.00 USD/EACH
Qty:
Representative photo only Explosion proof Three phase NEMA Type C face Motor 1 4 Hp 1800 RPM 208 230 460 VAC
Explosion-proof Three-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM, 208-230/460 VAC
Product Type Three-phase explosion-proof motors,hp 1/4,Model 56T17E5310/ G827
EW-71007-05
Usually ships in 10 days.
$545.00 USD/EACH
Qty:
Representative photo only Explosion proof Three phase NEMA Type C face Motor 3 4 Hp 208 230 460 VAC
Explosion-proof Three-phase NEMA Type C-face Motor, 3/4 Hp, 208-230/460 VAC
Product Type Three-phase explosion-proof motors,hp 3/4,Model 56T17G5316/ G840
EW-71007-35
Usually ships in 10 days.
$636.00 USD/EACH
Qty:
Representative photo only Explosion proof Single phase NEMA Type C face Motor 1 4 Hp 1800 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 1/4,Model G852
EW-71006-05
Usually ships in 10 days.
$489.00 USD/EACH
Qty:
Representative photo only Explosion proof Three phase NEMA Type C face Motor 1 3 Hp 1800 RPM 208 230 460 VAC
Explosion-proof Three-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM, 208-230/460 VAC
Product Type Three-phase explosion-proof motors,hp 1/3,Model 56T17E5311/ G828
EW-71007-15
In stock.
$573.00 USD/EACH
Qty:
Representative photo only Explosion proof Single phase NEMA Type C face Motor 1 3 Hp 1800 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 1/3,Model G855
EW-71006-15
Usually ships in 10 days.
$604.00 USD/EACH
Qty:
Representative photo only Explosion proof Single phase NEMA Type C face Motor 3 4 Hp 1800 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 3/4 Hp, 1800 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 3/4,Model G857
EW-71006-35
Usually ships in 10 days.
$676.00 USD/EACH
Qty:
Representative photo only Explosion proof Single phase NEMA Type C face Motor 1 2 Hp 3600 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 1/2 Hp, 3600 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 1/2,Model G854
EW-71006-20
Usually ships in 10 days.
$498.00 USD/EACH
Qty:
Representative photo only Explosion proof Single phase NEMA Type C face Motor 3 4 Hp 3600 RPM 115 208 230 VAC
Explosion-proof Single-phase NEMA Type C-face Motor, 3/4 Hp, 3600 RPM, 115/208-230 VAC
Product Type Single-phase explosion-proof motors,hp 3/4,Model G856
EW-71006-30
Usually ships in 10 days.
$762.00 USD/EACH
Qty:
Representative photo only Explosion proof Three phase NEMA Type C face Motor 1 Hp 1800 RPM 208 230 460 VAC
Explosion-proof Three-phase NEMA Type C-face Motor, 1 Hp, 1800 RPM, 208-230/460 VAC
Product Type Three-phase explosion-proof motors,hp 1,Model 56T17G5317/ G841
EW-71007-45
In stock.
$645.00 USD/EACH
Qty:
12 results (showing 1 - 12 )
Per Page 
Home  >  Pumps > Motors > AC Single and Three Motors for Positive Displacement Pumps > Explosion-Proof NEMA Type C-Face Motors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title