United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$575 to $650
(1)
$725 to $799
(1)
Decibel (dB) range (low):
33
(1)
30
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Sound Meters
> Extech Auto-Ranging Sound Level Datalogger
Extech Auto-Ranging Sound Level Datalogger
Stores up to 128,000 sound measurements and downloads to your computer for analysis
Datalogger featuers an analog bar graph to visually indicate the sound level. Time stamp logs each data point with the time and date.
Compliance:
ANSI S1.4-1983 Type 2, IEC 651 Type 2
Order sound level calibrator separately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Extech Sound Level Datalogger
Product Type Sound Meter,Meter class Type 2,Decibel (dB) range high 130
EW-01619-14
Usually ships in 8 days.
$649.99 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Extech Auto-Ranging Sound Level Datalogger
Extech Sound Level Datalogger with NIST-traceable Certificate of Calibration
Product Type Sound Meter,Meter class Type 2,Decibel (dB) range high 130
EW-01619-10
Usually ships in 15 days.
$774.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Extech Auto-Ranging Sound Level Datalogger
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Sound Meters
> Extech Auto-Ranging Sound Level Datalogger
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title