United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Life Science
>
Forensics
>
Safety & Response Products
>
Air Sampling and Gas Analysis
>
Indoor Air Quality/IAQ
> Extech Indoor Air Quality Meter - Datalogger
Extech Indoor Air Quality Meter - Datalogger
Automatically logs up to 20,000 records or manually stores up to 99 sets
Triple-line display allows you to view multiple parameters at once
Check indoor air quality in office buildings, greenhouses, laboratories, and anywhere that levels of carbon dioxide is a concern. This meter checks carbon dioxide, temperature, humidity, dew point, and wet bulb. Maintenance-free dual-wavelength NDIR (nondispersive infrared) carbon dioxide sensor ensures stable, accurate readings. Calibrates easily in fresh air. Capture the CO
2
min/max with time stamp and set an audible alarm for high/low CO
2
levels. Meter also features data hold and an easy-to-use keypad. Manually store and recall up to 99 set of data; or set meter to continuous logging and capture up to 20,000 records for later download through the RS-232 interface.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Extech Indoor Air Quality Meter
Range CO
2
0 to 6000 ppm,Brand Extech,Model EA80
EW-10101-55
In stock.
$789.99 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Extech Indoor Air Quality Meter - Datalogger
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Life Science
>
Forensics
>
Safety & Response Products
>
Air Sampling and Gas Analysis
>
Indoor Air Quality/IAQ
> Extech Indoor Air Quality Meter - Datalogger
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title