Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Data Loggers and Chart Recorders
>
Humidity Data Loggers and Recorders
> Extech RH520A Temperature/Humidity Paperless Recorders
Extech RH520A Temperature/Humidity Paperless Recorders
Ideal for monitoring controlled environments such as food processing, cleanrooms, and storage areas.
Paperless recorder measures temperature, humidity plus calculates dew point
Internal memory records up to 49,000 data points
Unit features audible and visual alarms with hi/low setpoints
Easy-to-operate digital recorder monitors and records temperature and humidity data in laboratories, storage and cleanrooms, freezers, and other critical areas. The detachable temperature/humidity probe extends up to one meter—ideal for measurements in closed environments. Probe does not require any calibration.
The large dual-graphic LCD simultaneously displays temperature (°F/°C) and relative humidity readings, plus time, date, battery status, and % of remaining memory. Dew point is easily displayed by pressing the dew point button. LCD also features adjustable vertical and horizontal resolution. Internal memory records up to 49,000 data points which can be transferred to a PC via RS-232 serial port for further analysis.
Audible and visual alarms generate warnings when conditions are outside selected high and low limits. A push-button lockout security feature prevents unauthorized tampering with settings. Recorder fits on your benchtop or easily mount to the wall.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Extech RH520A Temperature/Humidity Paperless Recorder with LCD and Detachable Probe
Product Type Paperless Recorders,Min temperature º C -28,Min temperature º F -20
EW-23029-81
In stock.
$300.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Extech RH520A Temperature/Humidity Paperless Recorders
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Data Loggers and Chart Recorders
>
Humidity Data Loggers and Recorders
> Extech RH520A Temperature/Humidity Paperless Recorders
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity