Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Kimax (17)
Product Type:
Availability:
In Stock (18)
CE Compliance:
Yes (60)
Capacity (Liters):
0 to 1 (1)
Capacity (mL):
Class:
Diameter (mm):
Funnel size (mL):
250 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Extraction Apparatus

Extraction Apparatus

Extraction Apparatus

The Kjeldahl determination of nitrogen came as a result of beer. Danish chemist Johan Kjeldhal was hired by Copenhagen’s Carlsberg Laboratory in the 1870s. The lab was operated by Carlsberg Brewery which today is the fourth largest brewery in the world. The Danish beer company wanted to find a way to determine how much protein was in the grain used to make its malt. You got more beer if there was less protein. This is how Johan began researching his Kjeldahl determination. He focused on measuring the nitrogen in the protein. Now the multi-step Kjeldahl determination is used for protein analysis in the almost all foods and beverages. Cole-Parmer has an array of extraction and analytical glassware apparatuses for various applications, including: arsine, distillation, cyanide, Kjeldahl determination, Kuderna-Danish concentration, and solvent recovery. Outside of glassware, choose from various extraction heaters to address Kjeldahl determination on a macro or micro scale. Use soxhlet extraction glassware systems for continuous extraction of solids with a suitable solvent. If you have questions about the Kjeldahl determination or any other extraction process and supplies please contact one of our Technical Experts who will be happy to assist you.
 Product Detail
Sort
 Availability
 Price

Environmental Express Snip & Pour Lab Control Standards, 40mg HEM/10mL Ace.; 20/Pk

  • Product Type: Organic Chemicals
  • CAS Number: 67-64-1, 544-76-3, and 57-11-4
  • Form: Liquid
EW-35200-22
Usually ships in 4 days.
$105.03 USD/PACKAGE

Qty:

Environmental Express SimpleDist Collection Traps, 25 mL; 100/Pk

  • Product Type: Collection Trap
  • Material: Polystyrene
  • Capacity: 25 mL
EW-35209-90
Available for order
or call 1-888-358-4705 to order
$196.17 USD/PACKAGE

Qty:

Environmental Express TCLP Filters, Standard, 90mm, 0.7µm; 100/Pk

  • Material: Binderfree borosilicate glass fiber
  • Product Type: Filter Papers
  • Diameter: 90 mm
EW-35208-56
Available for order
or call 1-888-358-4705 to order
$69.98 USD/PACKAGE

Qty:

Glass Paley Bottles, Cheese Cream Test, 9 gram Sample; 3/cs

  • Product Type: Extraction bottles
  • Material: Glass
  • Qty/pk: 3
EW-34590-40
Available for order
or call 1-888-358-4705 to order
$225.00 USD/CASE of 3

Qty:

Environmental Express TCLP Filters, Acid Wash, 47mm, 0.7µm; 100/Pk

  • Material: Binderfree borosilicate glass fiber
  • Product Type: Filter Papers
  • Diameter: 47 mm
EW-35208-47
Usually ships in 7 days.
$83.98 USD/PACKAGE

Qty:

Kimax 586000-0023 Soxhlet Extractor body; 300 mL; 1/Pk

  • Product Type: Soxhlet extractor body
  • Volume: 300 mL
  • Joint size: Top: 55/50
    Bottom: 24/40
EW-99037-42
In stock.
$241.00 USD/EACH

Qty:

Environmental Express TCLP Filters, Standard, 142mm, 0.7µm; 100/Pk

  • Material: Binderfree borosilicate glass fiber
  • Product Type: Filter Papers
  • Diameter: 142 mm
EW-35208-59
In stock.
$169.98 USD/PACKAGE

Qty:

Mojonnier Glass Extraction Flask, 25 mL; 1/Pk

  • Capacity: 25 mL
  • Material: Glass
  • Qty/ea: 1
EW-34590-65
In stock.
$77.00 USD/EACH

Qty:

Kimax 456000-0022 Allihn Condenser; 250 mL; 1/Pk

  • Product Type: Soxhlet condensers
  • Volume: 250 mL
  • Condenser type: Allihn
EW-99037-28
Usually ships in 14 days.
$171.00 USD/EACH

Qty:

Glass Babcock Bottle, Milk Test, 18 gram Sample; 1/Pack

  • Product Type: Extraction bottles
  • Material: Glass
  • Qty/pk: 1
EW-34590-06
In stock.
$30.00 USD/EACH

Qty:

Electrothermal Electromantle Extraction Heater - 6 positions, 500 mL, 115V

  • Product Type: Extraction Heater
  • Watts: 1200
  • Flask size: 500 mL
EW-03076-30
Usually ships in 6 days.
$2,250.00 USD/EACH

Qty:

Kimax 586000-0022 Soxhlet Extractor body; 250 mL; 1/Pk

  • Product Type: Soxhlet extractor body
  • Volume: 250 mL
  • Joint size: Top: 45/50
    Bottom: 24/40
EW-99037-40
Available for order
or call 1-888-358-4705 to order
$224.00 USD/EACH

Qty:

Glass Babcock Bottles, Ice Cream Test, 18 gram Sample; 12/cs

  • Product Type: Extraction bottles
  • Material: Glass
  • Qty/cs: 12
EW-34590-10
Usually ships in 14 days.
$276.00 USD/CASE of 12

Qty:

Kimax 456250-0022 Friedrich Condenser; 250 mL; 1/Pk

  • Product Type: Soxhlet condensers
  • Volume: 250 mL
  • Condenser type: Freidrich
EW-99037-34
Usually ships in 14 days.
$226.00 USD/EACH

Qty:

Electrothermal MQ 6-Well Micro-Kjeldahl Extraction Heater, 800 mL, 115V US

  • Product Type: Extraction Heater
  • Watts: 3300
  • Flask max capacity: 800 mL
EW-03075-18
Available for order
or call 1-888-358-4705 to order
$4,108.00 USD/EACH

Qty:

319 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Extraction Apparatus