Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Eyewear and Safety Glasses
Eyewear and Safety Glasses
Eyewear and Safety Glasses
Introduction to Eye Protection
OSHA requirements state that protective eye and face devices must comply with ANSI Z87.1 (American Standard Protection). ANSI Z87.1 requires that all lenses be permanently imprinted with the manufacturer’s monogram and all frames marked ”Z87”.
Types of eyewear
Safety glasses have impact-resistant lenses and strong frames to protect you from flying particles.
Safety goggles are designed to provide a shield around the entire eye area to protect against hazards from many directions.
Face shields give you full face protection against sparks, splashes, and splatters. They provide secondary protection and must be worn with protective glasses or goggles.
 Product Detail
Sort
 Availability
 Price
Safety Eyewear Organizer, Acrylic, 12 Slots, Wall-Mount/Benchtop
  • Product Type: Glass Holder
  • Capacity: 12 pair
  • Dimensions: 12-1/4 " W
EW-86307-00
Available for order
or call 1-888-358-4705 to order
$108.00 USD/EACH
Qty:
3M Virtua Safety Eyewear, Clear frame, Clear lens, Anti-fog
  • Product Type: Glasses
  • Color: Clear
  • Lens: Clear
EW-86472-74
In stock.
$3.50 USD/EACH
Qty:
Sellstrom Acetate Face Shield with Headgear, Clear Anti-Fog Shield
  • Product Type: Face Shields
  • Color: Blue
  • Lens: Clear
EW-86478-17
In stock.
$36.75 USD/EACH
Qty:
Hilco 70 Oc-Clear Lens Cleaner Towelettes; 100/Pk
  • Product Type: Cleaning Supplies
  • Qty/pk: 100
  • Brand: Hilco
EW-86212-00
In stock.
$10.75 USD/PACKAGE of 100
Qty:
3M Virtua Sport Safety Eyewear, Clear frame, Clear lens
  • Product Type: Glasses
  • Color: Clear
  • Lens: Clear
EW-86472-01
In stock.
$4.00 USD/EACH
Qty:
Sellstrom Odyssey II Autoclavable Safety Goggles, Top vented
  • Product Type: Safety Goggles
  • Color: White
  • Lens: Clear
EW-86281-02
In stock.
$14.00 USD/EACH
Qty:
Uvex by Honeywell Stealth Safety Goggles, One Size, Gray Body/Clear Lens
  • Product Type: Safety Goggles
  • Color: Gray
  • Lens: Clear
EW-86326-00
In stock.
$18.75 USD/EACH
Qty:
3M BX Dual Reader Safety Glasses, 1.5x top and bottom diopters
  • Product Type: Glasses
  • Color: Black and Silver
  • Lens: Clear
EW-86474-31
In stock.
$16.75 USD/EACH
Qty:
North by Honeywell Protecto-Shield Adjustable Head Gear, Black HDPE
  • Product Type: Face Shields
  • Color: Black and Clear
  • Lens: Clear
EW-81690-00
In stock.
$23.25 USD/EACH
Qty:
3M BX Dual Reader Safety Glasses, 2.0X top and bottom diopters
  • Product Type: Glasses
  • Color: Black and Silver
  • Lens: Clear
EW-86474-33
In stock.
$16.75 USD/EACH
Qty:
Uvex by Honeywell Slate Safety Glasses, Gunmetal, Antifog/Clear Lens
  • Product Type: Glasses
  • Color: Gunmetal
  • Lens: Clear
EW-86321-61
In stock.
$14.75 USD/EACH
Qty:
Tabletop or Wall Mount Safety Goggles/Glasses Dispenser
  • Product Type: Organizer
  • Color: Clear
  • Dimensions: 8 " W
EW-86456-20
In stock.
$86.00 USD/EACH
Qty:
Safety Eyewear Organizer, Acrylic, 20 Slots, Wall-Mount/Benchtop
  • Product Type: Glass Holder
  • Capacity: 20 pair
  • Dimensions: 15-1/4 " W
EW-86307-05
In stock.
$127.00 USD/EACH
Qty:
Smith & Wesson 19794 Magnum Glasses, Fog-free Clear Lens
  • Product Type: Glasses
  • Color: Black
  • Lens: Clear/Anti-fog
EW-86285-01
In stock.
$12.75 USD/EACH
Qty:
Allsafe SMC MonoShield Combination Googles/Face Shield, Clear Anti-Fog
  • Product Type: Face Shields
  • Color: Clear
  • Lens: Clear/Anti-fog
EW-81657-30
In stock.
$44.00 USD/EACH
Qty:
120 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Eyewear and Safety Glasses