log in or register
Product Type:
Availability:
Power (VAC):
120 (6)
Power (VDC):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Alarms > Federal Signal Low Profile Strobe Light
Federal Signal Low Profile Strobe Light
Federal Signal Low Profile Strobe Light
  • Long life in a compact strobe light
NEMA 4X/IP65-rated, low-profile strobes are ideal for mounting on control panels and other flat or flush surfaces. Strobes operate in temperatures from -31 to 150°F (-35 to 66°C). Threaded screw-on lens provides tool-free wiring and strobe tube replacement—strobe tube rated for 7000 hours of use. High-quality, long life strobe lamps are designed to reduce tungsten build-up for longer lamp maintenance cycles. Strobe mechanism produces 2.2 joules of energy while drawing low amperage. Strobes have a flash rate of 75 to 85 flashes per minute. Availablein three mounting configurations: 1/2" NPT(M) pipe mount with 18" wire leads, 1/2" NPT(F) pipe mount, and 3-hole surface mount.
 Product Detail
Sort
 Availability
 Price
Representative photo only Federal Signal Low Profile Strobe Light Surface mount Amber
Federal Signal Low Profile Strobe Light, Surface mount , Amber
Product Type Strobes - surface-mount,Power VAC 120,Brand Federal Signal
EW-10402-35
Usually ships in 5 days.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light Surface mount Amber
Federal Signal Low Profile Strobe Light, Surface mount , Amber
Product Type Strobes - surface-mount,Power VDC 12 to 48,Brand Federal Signal
EW-10402-30
Usually ships in 12 days.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light Surface mount Red
Federal Signal Low Profile Strobe Light, Surface mount , Red
Product Type Strobes - surface-mount,Power VDC 12 to 48,Brand Federal Signal
EW-10402-34
Usually ships in 12 days.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light Surface mount Red
Federal Signal Low Profile Strobe Light, Surface mount , Red
Product Type Strobes - surface-mount,Power VAC 120,Brand Federal Signal
EW-10402-39
Call 1-888-358-4705 for availability.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light NPT M pipe mount Amber
Federal Signal Low Profile Strobe Light, ½" NPT(M) pipe mount, Amber
Product Type Strobes - pipe-mount,Power VAC 120,Connections 1/2" NPT(M)
EW-10402-05
Usually ships in 3 days.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light Surface mount Green
Federal Signal Low Profile Strobe Light, Surface mount , Green
Product Type Strobes - surface-mount,Power VAC 120,Brand Federal Signal
EW-10402-38
In stock.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light NPT F pipe mount Red
Federal Signal Low Profile Strobe Light, ½" NPT(F) pipe mount , Red
Product Type Strobes - pipe-mount,Power VAC 120,Connections 1/2" NPT(F)
EW-10402-24
Usually ships in 7 days.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light NPT F pipe mount Red
Federal Signal Low Profile Strobe Light, ½" NPT(F) pipe mount , Red
Product Type Strobes - pipe-mount,Power VDC 12 to 48,Connections 1/2" NPT(F)
EW-10402-19
Usually ships in 10 days.
$69.86 USD/EACH
Qty:
Representative photo only Federal Signal Low Profile Strobe Light NPT M pipe mount Red
Federal Signal Low Profile Strobe Light, ½" NPT(M) pipe mount, Red
Product Type Strobes - pipe-mount,Power VAC 120,Connections 1/2" NPT(M)
EW-10402-09
Usually ships in 12 days.
$69.86 USD/EACH
Qty:
9 results (showing 1 - 9 )
Per Page 
Home  >  Safety Products > Alarms > Federal Signal Low Profile Strobe Light
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title