Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Availability:
Capacity (mL):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Laboratory Supplies & Consumables > Shippers > Fiberboard Mailers > Fiberboard Mailer with Boston round glass bottle

Fiberboard Mailer with Boston round glass bottle

Fiberboard Mailer with Boston round glass bottle

  • Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes—each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.
 Product Detail
Sort
 Availability
 Price
Representative photo only Boston Bottle Mailer 8 oz 6 1 2 L x 2 1 2 dia 24 cs

Boston Bottle Mailer, 8 oz, 6-1/2" L x 2-1/2" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.237
EW-06148-42
In stock.
$180.00 USD/CASE

Qty:

Representative photo only Boston Bottle Mailer 2 oz 4 1 2 L x 1 3 4 dia 24 cs

Boston Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.059
EW-06148-22
Usually ships in 10 days.
$154.00 USD/CASE

Qty:

Representative photo only Boston Bottle Mailer 4 oz 5 1 2 L x 2 dia 24 cs

Boston Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.125
EW-06148-32
Usually ships in 10 days.
$208.00 USD/CASE

Qty:

Representative photo only Boston Round Glass Bottle Mailer 1 oz 3 3 4 L x 1 1 2 dia 24 cs

Boston Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.03
EW-06148-12
Usually ships in 10 days.
$124.00 USD/CASE

Qty:

4 results (showing 1 - 4 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Shippers > Fiberboard Mailers > Fiberboard Mailer with Boston round glass bottle

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title