Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Availability:
Capacity (mL):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Laboratory Supplies & Consumables > Shippers > Fiberboard Mailers > Fiberboard Mailing Tubes

Fiberboard Mailing Tubes

Fiberboard Mailing Tubes

  • Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes—each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps.
 Product Detail
Sort
 Availability
 Price
Representative photo only Bottle Mailer 4 oz 5 1 2 L x 2 dia 24 cs

Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.125
EW-06148-31
In stock.
$125.00 USD/CASE

Qty:

Representative photo only Bottle Mailer 2 oz 4 1 2 L x 1 3 4 dia 24 cs

Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.06
EW-06148-21
Call 1-888-358-4705 for availability.
$104.00 USD/CASE

Qty:

EW-06148-11 Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs

Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs

Product Type Mailers,Material Fiberboard,Capacity Liters 0.03
EW-06148-11
Usually ships in 3 days.
$123.00 USD/CASE

Qty:

3 results (showing 1 - 3 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Shippers > Fiberboard Mailers > Fiberboard Mailing Tubes

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title