Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Advantec (545)
Whatman (162)
Product Type:
Availability:
In Stock (509)
Diameter (mm):
Housing material:
Acrylic (29)
Capacity (mL):
0 - 20 (2)
Flow rate:
20 GPM (21)
40 GPM (21)
35 GPM (16)
Cartridge size:
Connection:
Funnel size (mL):
300 (13)
500 (11)
1000 (10)
250 (5)
100 (4)
15 (4)
150 (4)
13/100 (1)
Diameter:
50 mm (9)
70 mm (8)
90 mm (6)
Capacity (gal.):
1-5 (6)
Capacity (Liters):
0 to 1 (4)
1 to 5 (2)
Effective filtration area:

Learn more about
Filtration Technology

2,600 pages
of answers

FREE
2013/14 Cole-Parmer
Sourcebook
Over 40 pages of filtration housing products including hundreds of pages of flow accessories such as fittings, tubing and process control.
Request your copy today.

Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Filtration

Types of filter housings available at Cole-Parmer

Bottle-Top Filters;

Bottle Top Filters

  • Direct filtration into sterile glass media bottles
  • Vacuum port in middle juncture vacuum allows for faster filtration due to pressure differential at filter membrane versus gravity feed

Nalgene Disposable S

Filter Units

  • Membrane holder, vacuum arm and receivers are complete system
  • Filter application for Cell Culture Media, Biological Fluids, Aqueous Solutions

Disposable In-Line Gas/Liquid Filters

In-line Filter Holders

  • Available with connections for tubing, pipes and syringes
  • Range of materials for suitability with applications such as liquids or gas filtration for acid, oil, high purity compressed gas and biological media

Millipore Microcon<r

Centrifuge Filters

  • More effective filtration due to extreme pressure at filtration membrane from centrifugal forces.
  • Advantages highly reproducible, minimal sample loss
  • Applications sample concentration, protein, peptide, DNA purification, buffer exchange, salt & radio label removal

Glass Fiber Prefilters with Final Nonsterile Cellu

Syringe Filters

  • Bi-directional filters
  • Available with luer slip and lock fit connections
  • Applications HPLC, microbiological analysis, waste water treatment, sterilization of aqueous solutions, cryogenic fluids, macromolecules, protein binding

Glass Fiber Prefilters with Final Nonsterile Cellu

Vacuum filtration Holders and Manifolds

  • Connect 3-6 filter holders to one vacuum source. 2-3 way valves available to control vacuum or venting.
  • Application ideal for testing of antibiotics and pharmaceuticals b bacteria retentive methods

Polypropylene bag fi

Bag Filters

  • High flow up to 150 LPM
  • Large solids 1 to 800 m
  • Pressure ratings up to 6.8 bar
  • Range of compatibility with organic solvents, animal fat, petroleum, vegetable oils, micro-organisms, alkalis, organic acids, oxidizing agents and mineral acids
  • Available with pressure gauge

Hayward <small><sup>�</sup></small>  single-

Heavy duty bag filter

  • High flow up to 830 LPM
  • Available with a wide range of bags for temperature, flow rate and filtration quality
  • Offer 98% efficiency at rated pore size
  • Easy bag change out
  • Applications filtration of coolants, solvents, process water, ground water, oils for foods, lubrication and hydraulics

Double open end Slim

Cartridge style filters

  • Housing sizes up to 20" in metal and plastic designs for standard and high purity applications
  • Temperatures up to 157°C
  • Pressures up to 17 bar
  • Stacked systems up to 20 double open end cartridges in one housing

Cartridges types
Double open end cartridges

  • Over 50 items with standard diameter of 2.75" for sediment, organic, lead, scale, and deionization treatment
  • Filtration down to 0.2 micro with flows up to 76 LPM
  • Includes variations that are autoclavable, biologically safe and FDA approved

High flow double open end cartridges

  • 4.5" diameter for sediment, organiscs, oil, iron and deionization treatment
  • Filtration down to 0.5 microns with flows up to 190 LPM

Single open end cartridges

  • Includes antimicrobial elements with filtration down to 0.1 micron
  • Cartridges meet USP, FDA and NSF criteria
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only Advantec Mixed Cellulose Ester Membranes 47 mm dia 0 45 m 100 pack

Advantec Mixed Cellulose Ester Membranes; 47 mm dia, 0.45 µm; 100/pack

Product Type Mixed Cellulose Ester Membranes,Material Mixed Cellulose Ester,Flow rate water 0.045 L/min/cm2
EW-06639-14
In stock.
$123.00 USD/PACKAGE of 100

Qty:

Representative photo only Advantec Polypropylene Filter Holder for 47 mm membranes

Advantec Polypropylene Filter Holder for 47-mm membranes

Product Type Polypropylene Filter Holders,Product Type Polypropylene Filter Holders,Material Polypropylene
EW-06623-22
In stock.
$58.00 USD/EACH

Qty:

Representative photo only Cole Parmer Nylon Nonsterile Syringe Filters 0 45 pore size 25 mm diameter

Cole-Parmer Nylon Nonsterile Syringe Filters; 0.45 µ pore size; 25 mm diameter

Product Type Syringe Filters,Housing material Acrylic,Color Natural
EW-02915-14
In stock.
$180.00 USD/BOX of 100

Qty:

EW-06730-04 Thermo Scientific Nalgene Disposable Sterile Filter Units; membrane, grided, CN; capacity, 150mL; pore size, 0.45&micro;; 12/pack

Thermo Scientific Nalgene Disposable Sterile Filter Units; membrane, grided, CN; capacity, 150mL; pore size, 0.45; 12/pack

Product Type Cellulose Nitrate Filter Units,Housing material Polypropylene Upper; High-Impact Polystyrene Lower,Color White
EW-06730-04
In stock.
$110.00 USD/DOZEN

Qty:

Representative photo only Cole Parmer PTFE Nonsterile Syringe Filters 0 2

Cole-Parmer PTFE Nonsterile Syringe Filters; 0.2µ

Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-02915-28
In stock.
$95.00 USD/BOX of 20

Qty:

Representative photo only Cole Parmer PTFE Nonsterile Syringe Filters 0 45

Cole-Parmer PTFE Nonsterile Syringe Filters; 0.45µ

Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-02915-22
Call 1-888-358-4705 for availability.
$180.00 USD/BOX of 100

Qty:

Representative photo only Thermo Scientific Polyethersulfone PES Filter Units membrane PES capacity 500mL pore size 0 20 12 pack

Thermo Scientific Polyethersulfone (PES) Filter Units; membrane; PES; capacity, 500mL; pore size, 0.20µ; 12/pack

Product Type Polyethersulfone Filter Units,Sterile Yes,Membrane material Polyethersulfone (PES)
EW-06730-43
In stock.
$210.00 USD/PACKAGE

Qty:

EW-02915-20 Cole-Parmer<small><sup>&reg;</sup></small> PTFE Nonsterile Syringe Filters; 0.20&micro;

Cole-Parmer PTFE Nonsterile Syringe Filters; 0.20

Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-02915-20
In stock.
$180.00 USD/BOX of 100

Qty:
See all PTFE Filters

Representative photo only Thermo Scientific Polyethersulfone PES Filter Units 150 mL 0 20 m Pore Size

Thermo Scientific Polyethersulfone (PES) Filter Units; 150 mL, 0.20 µm Pore Size

Product Type Polyethersulfone Filter Units,Sterile Yes,Membrane material Polyethersulfone (PES)
EW-06730-31
Usually ships in 8 days.
$92.00 USD/PACKAGE

Qty:

Representative photo only Cole Parmer PTFE Nonsterile Syringe Filters pore size 0 45

Cole-Parmer PTFE Nonsterile Syringe Filters; pore size; 0.45µ

Product Type Syringe Filters,Housing material Polypropylene,Color Natural
EW-29550-10
In stock.
$135.00 USD/PACKAGE of 100

Qty:

EW-29830-00 Double open end high-efficiency pleated polypropylene filter cartridge; 0.2 &micro;m, 10"L

Double open end high-efficiency pleated polypropylene filter cartridge; 0.2 m, 10"L

Product Type Filter Cartridges,Material Polypropylene,Filter size " L 10
EW-29830-00
In stock.
$94.00 USD/EACH

Qty:

EW-34509-00 Labglass<small><sup>&#153;</sup></small> Filtration Assembly, 47mm dia, 300 mL

Labglass Filtration Assembly, 47mm dia, 300 mL

Product Type Filtration Assemblies,Diameter mm 47,Qty/pk 1
EW-34509-00
In stock.
$185.00 USD/CASE

Qty:

Representative photo only Advantec Sterile Mixed Cellulose Ester Filter Membranes 47mm dia 0 45m absorbent pads not included gridded surface 1000 pack packaged individually

Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.45µm; absorbent pads not included; gridded surface; 1000/pack (packaged individually)

Product Type Mixed Cellulose Ester Membranes,Material Mixed Cellulose Ester, White Membrane, individually packaged,Diameter mm 47
EW-06644-31
In stock.
$355.00 USD/PACKAGE

Qty:

Representative photo only Disposable In Line Gas Liquid Filters Nylon gas efficiency 93 liquid efficiency 25 5 pack

Disposable In-Line Gas/Liquid Filters; Nylon; gas efficiency, 93%; liquid efficiency, 25µ; 5/pack

Product Type Gas/Liquid Filters, In-Line,Housing material Nylon,Max pressure psi 125
EW-02909-10
In stock.
$145.00 USD/PACKAGE of 5

Qty:

Representative photo only Double open end semiconductor filter cartridge 0 20 m

Double open end semiconductor filter cartridge; 0.20 µm

Product Type Filter Cartridges, Double-Open-End,Length in. 10,Pore size µm 0.2
EW-06479-60
In stock.
$135.00 USD/EACH

Qty:

1794 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title