log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Flare Fittings > Flare Tee
Flare Tee
Flare Tee
  • Leak tight & high-pressure connection for corrosive chemical applications
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leak-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing. PFA has a low coefficient of friction and therefore does not form a tight seal with standard gripper fittings. Flare fittings are a great alternative to gripper fittings in critical applications. To install Flare fittings you slide the flare nut on to a piece of PFA tubing, the tubing is then heated using a heat gun and then a mandrel is inserted in the fitting to create the flare. The flared end of the tubing is then inserted into the body of the fitting and the nut tightened down on the flare. Choose from the various styles. If there is another type or style not show call our Application Specialists for more information on addition fitting types.
 Product Detail
Sort
 Availability
 Price
EW-30490-84 Flare union Tee, 1/2", PTFE, Each
Flare union Tee, 1/2", PTFE, Each
Product Type Compression fittings,Material PTFE,Brand Parker Hannifin
EW-30490-84
Call 1-888-358-4705 for availability.
$94.00 USD/EACH
Qty:
See all Flare Tee
Representative photo only Flare union Tee 1 4 PTFE Each
Flare union Tee, 1/4", PTFE, Each
Product Type Compression fittings,Material PTFE,Brand Parker Hannifin
EW-30490-80
Call 1-888-358-4705 for availability.
$67.00 USD/EACH
Qty:
See all Flare Tee
Representative photo only Flare union Tee 3 4 PTFE Each
Flare union Tee, 3/4", PTFE, Each
Product Type Compression fittings,Material PTFE,Brand Parker Hannifin
EW-30490-86
Call 1-888-358-4705 for availability.
$135.00 USD/EACH
Qty:
See all Flare Tee
Representative photo only Flare union Tee 3 8 PTFE Each
Flare union Tee, 3/8", PTFE, Each
Product Type Compression fittings,Material PTFE,Brand Parker Hannifin
EW-30490-82
Call 1-888-358-4705 for availability.
$74.00 USD/EACH
Qty:
See all Flare Tee
4 results (showing 1 - 4 )
Per Page 
Home  >  Fittings > Flare Fittings > Flare Tee
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title