log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Flare Fittings > Flaring Tools for PFA Flare Fittings
Flaring Tools for PFA Flare Fittings
Flaring Tools for PFA Flare Fittings
Use these tools for a better fit. Heat the PFA tubing with a heat gun, and insert the tool to create the flare. Tools are available for any size of tubing from 1/4-inch to 1-inch outer diameter.
 Product Detail
Sort
 Availability
 Price
EW-30490-01 Flaring tool for tubing 1/4 to 1" outer diameter
Flaring tool for tubing 1/4 to 1" outer diameter
Product Type Compression fittings,Brand Parker Hannifin
EW-30490-01
Call 1-888-358-4705 for availability.
$360.00 USD/EACH
Qty:
Representative photo only Flaring tool for tubing 1 4 outer diameter
Flaring tool for tubing 1/4" outer diameter
Product Type Compression fittings,Brand Parker Hannifin
EW-30490-03
Call 1-888-358-4705 for availability.
$115.00 USD/EACH
Qty:
Representative photo only Flaring tool for tubing 3 8 outer diameter
Flaring tool for tubing 3/8" outer diameter
Product Type Compression fittings,Brand Parker Hannifin
EW-30490-05
Call 1-888-358-4705 for availability.
$115.00 USD/EACH
Qty:
Representative photo only Flaring tool for tubing 1 2 outer diameter
Flaring tool for tubing 1/2" outer diameter
Product Type Compression fittings,Brand Parker Hannifin
EW-30490-07
Call 1-888-358-4705 for availability.
$115.00 USD/EACH
Qty:
Representative photo only Flaring tool for tubing 3 4 outer diameter
Flaring tool for tubing 3/4" outer diameter
Product Type Compression fittings,Brand Parker Hannifin
EW-30490-09
Call 1-888-358-4705 for availability.
$115.00 USD/EACH
Qty:
5 results (showing 1 - 5 )
Per Page 
Home  >  Fittings > Flare Fittings > Flaring Tools for PFA Flare Fittings
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title