log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Flexible PVC Fittings
Flexible PVC Fittings
Flexible PVC Fittings
  • Introduce the element of flexibility to rigid tubing systems
  • Absorb shock and accommodate vibration
  • Excellent angular flexibility – up to 180deg!
These fittings provide flexibility for expansion and contraction in rigid tubing systems. For use with any tubing material – including glass, polycarbonate, and metal – simply slide the tubing end into the fitting for a friction fit. If desired, use adhesive or clamps to enhance the security of the fitting-to-tubing connection.
Made of a soft, flexible PVC material, these fittings withstand a wide variety of chemicals, especially strong oxidizing agents, reducing agents, and mineral oils. Usable for temperatures from -41 to 165degF (-41 to 74degC). The PVC resin complies with FDA regulations for contact with food (21 CFR 175.300, 178.2650 and 178.3790) and are 3A approved by the dairy industry standards for fluids containing milk fat. Available in seven different configurations
 Product Detail
Sort
 Availability
 Price
EW-31545-02 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Elbow, 7/16" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-02
In stock.
$7.75 USD/EACH
Qty:
EW-31545-05 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Elbow, 3/4" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-05
Usually ships in 7 days.
$9.25 USD/EACH
Qty:
EW-31545-55 Image is for illustrative purposes only. Please refer to product description
Flexible PVC T Connector, 3/4" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-55
Usually ships in 7 days.
$23.50 USD/EACH
Qty:
EW-31545-45 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Y Connector, 3/4" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-45
Usually ships in 7 days.
$23.50 USD/EACH
Qty:
EW-31545-25 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Reducer Connector, 3/4 X 1/2" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-25
Usually ships in 7 days.
$6.25 USD/EACH
Qty:
EW-31545-35 Flexible PVC Straight Connector, 3/4" Tubing OD
Flexible PVC Straight Connector, 3/4" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-35
Usually ships in 7 days.
$8.25 USD/EACH
Qty:
EW-31545-26 Flexible PVC Reducer Connector, 7/8 X 5/8" Tubing OD
Flexible PVC Reducer Connector, 7/8 X 5/8" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-26
Usually ships in 7 days.
$6.75 USD/EACH
Qty:
EW-31545-24 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Reducer Connector, 5/8 X 7/16" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-24
Call 1-888-358-4705 for availability.
$5.75 USD/EACH
Qty:
EW-31545-06 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Elbow, 7/8" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-06
Usually ships in 7 days.
$10.50 USD/EACH
Qty:
EW-31545-22 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Reducer Connector, 5/16 X 7/16" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-22
In stock.
$5.00 USD/EACH
Qty:
EW-31545-03 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Elbow, 1/2" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-03
In stock.
$8.00 USD/EACH
Qty:
EW-31545-32 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Straight Connector, 7/16" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-32
Usually ships in 7 days.
$7.25 USD/EACH
Qty:
EW-31545-47 Flexible PVC Y Connector, 1" Tubing OD
Flexible PVC Y Connector, 1" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-47
Usually ships in 7 days.
$32.50 USD/EACH
Qty:
EW-31545-46 Image is for illustrative purposes only. Please refer to product description
Flexible PVC Y Connector, 7/8" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-46
In stock.
$27.00 USD/EACH
Qty:
EW-31545-14 Image is for illustrative purposes only. Please refer to product description
Flexible PVC 180° U-bend, 5/8" Tubing OD
Product Type Tubing fittings,Material PVC
EW-31545-14
Usually ships in 7 days.
$9.00 USD/EACH
Qty:
41 results (showing 1 - 15 )
Per Page 
Home  >  Fittings > Flexible PVC Fittings
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title