Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Clean Room
>
Airflow Measurement
> FlowMarker Fog Generator
FlowMarker Fog Generator
Visualize natural air currents without creating disturbances
Portable design allows operation in hard-to-reach areas
Nontoxic fog breaks down into carbon dioxide and water
Identify potential dead spots in room airflow patternsthe FlowMarker marks air movements through critical environment areas, fume hoods, biosafety cabinets, HVAC systems and low air velocity applications. The fog consists of FDA-approved, non-toxic polyfunctional alcohol preparation, which meets standards for USP purity (Food grade), and highly purified demineralized water. When full, the 16-mL fluid reservoir allows operation for 20 continuous minutes; order refill fluid below. Automatic shut-off after four minutes preserves battery life.
What’s included:
flow tube, rechargeable battery, battery charger/power supply, one fog refill can, and carrying case.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
FlowMarker Fog Generator
Product Type Fog Generator,Brand Degree Controls,Model FLOWMARKER KIT
EW-10322-30
Usually ships in 8 days.
$6,329.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
FlowMarker Fog Generator
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Clean Room
>
Airflow Measurement
> FlowMarker Fog Generator
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity