Home  >  Flow, Level & Valves > Flowmeters > Flow Controllers
Flow Controllers
Flow Controllers
Cole-Parmer offers an extensive range of flow meters and flow controllers for both liquid and gas applications. Cole-Parmer specializes in flow control of media in pipe lines less than 2" and offers flow metering solutions for measuring liquids flow from 0.0002 ml/min to over 4,000 LPM and for gases from 0.02 ml/min to over 500 m3/hour.
Cole-Parmer's flow meters come with a wide range of output options including set-point switches, analogue outputs 4-20 mA, 0-5 VDC, pulse signals and RS-232 options. Use Cole-Parmer's flow meter quick search to find the right flow meter for your application. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve your application needs.
Flow Meter Selection Considerations
When selecting a flow meter consideration must be given to:
  • Flow measurement type - momentum (velocity), volumetric or mass flow measurement
  • Media - type of media (liquid, gas or slurry) and any special condition such as particulates in the media and viscosity of the media
  • Media conditions - pressure and temperature of media and whether media conditions are likely to remain constant or vary
  • Flow range - required flow range of media (min and max readings required)
  • Accuracy - required accuracy of the readings
  • Environmental considerations - special installation considerations such as hygienic installation, installing in to an ATEX zone or requiring tamper proof readings
For special or unique flow control applications
Use our Flowmeter and Flow Meters Controller Application Table to narrow your selection.
 Product Detail
Sort
 Availability
 Price
EW-05631-00 Digital Flow Transmitter and Display, +GF+ Signet Loop Powered (7-30 VDC)
Digital Flow Transmitter and Display, +GF+ Signet Loop Powered (7-30 VDC)
Product Type Paddle Wheel Flowmeter,Output 4 to 20 mA, scalable,CE Compliance Yes
EW-05631-00
In stock.
$547.00 USD/EACH
Qty:
EW-05628-00 Batch Controller, 12-24 VAC/dc
Batch Controller, 12-24 VAC/dc
Product Type Paddle Wheel Flowmeter,Max operating temperature º F 131,Max operating temperature º C 55
EW-05628-00
In stock.
$1,064.00 USD/EACH
Qty:
EW-33112-50 Cole-Parmer<small><sup>&reg;</sup></small> flow rate/totalilzer display
Cole-Parmer flow rate/totalilzer display
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 158
EW-33112-50
Call 1-888-358-4705 for availability.
$506.00 USD/EACH
Qty:
EW-33110-60 New Flow Controllers
Flow Controllers, Panel Mount
Product Type Flow Controller,Max operating temperature º F 158,Max operating temperature º C 70
EW-33110-60
Call 1-888-358-4705 for availability.
$500.00 USD/EACH
Qty:
EW-33125-10 Paddle Wheel Flow Meter, Stainless Steel, 0.1 to 10 GPM
Paddle Wheel Flow Meter, Stainless Steel, 0.1 to 10 GPM
Product Type Flow Sensor,Max operating temperature º F 200,Max operating temperature º C 93
EW-33125-10
Usually ships in 10 days.
$766.00 USD/EACH
Qty:
EW-33110-05 Flow Sensor, TFE, 0.07 to 5 GPM
Flow Sensor, TFE, 0.07 to 5 GPM
Product Type Flow Sensor,Max operating temperature º F 250,Max operating temperature º C 121
EW-33110-05
In stock.
$628.00 USD/EACH
Qty:
EW-05629-22 3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
Product Type Paddle Wheel Flowmeter,Max operating temperature º F 131,Max operating temperature º C 55
EW-05629-22
Usually ships in 14 days.
$617.00 USD/EACH
Qty:
Representative photo only Flow Sensor PP 0 07 to 5 GPM
Flow Sensor, PP, 0.07 to 5 GPM
Product Type Flow Sensor,Max operating temperature º F 160,Max operating temperature º C 71
EW-33110-00
Call 1-888-358-4705 for availability.
$299.00 USD/EACH
Qty:
Representative photo only Flow Sensor PP 0 5 to 40 GPM
Flow Sensor, PP, 0.5 to 40 GPM
Product Type Flow Sensor,Max operating temperature º F 160,Max operating temperature º C 71
EW-33110-30
Usually ships in 7 days.
$299.00 USD/EACH
Qty:
Representative photo only Flow Sensor PP 0 2 to 20 GPM
Flow Sensor, PP, 0.2 to 20 GPM
Product Type Flow Sensor,Max operating temperature º F 160,Max operating temperature º C 71
EW-33110-20
Usually ships in 13 days.
$299.00 USD/EACH
Qty:
Representative photo only Flow Sensor TFE 0 1 to 10 GPM
Flow Sensor, TFE, 0.1 to 10 GPM
Product Type Flow Sensor,Max operating temperature º F 250,Max operating temperature º C 121
EW-33110-15
Call 1-888-358-4705 for availability.
$628.00 USD/EACH
Qty:
Representative photo only Flow Sensor PTFE 0 5 to 40 GPM
Flow Sensor, PTFE, 0.5 to 40 GPM
Product Type Flow Sensor,Max operating temperature º F 250,Max operating temperature º C 121
EW-33110-35
Usually ships in 15 days.
$628.00 USD/EACH
Qty:
EW-33110-70 New Flow Controllers
Flow Controllers, Wall-Mount
Product Type Flow Controller,Max operating temperature º F 158,Max operating temperature º C 70
EW-33110-70
Usually ships in 14 days.
$500.00 USD/EACH
Qty:
Representative photo only Flow rate totalizer analog input
Flow rate/totalizer, analog input
Product Type Flow Controller,Max operating temperature º F 122,Max operating temperature º C 50
EW-33114-10
Call 1-888-358-4705 for availability.
$460.00 USD/EACH
Qty:
EW-33114-00 Flow rate/totalizer, pulse input
Flow rate/totalizer, pulse input
Product Type Flow Controller,Max operating temperature º F 122,Max operating temperature º C 50
EW-33114-00
Call 1-888-358-4705 for availability.
$442.00 USD/EACH
Qty:
27 results (showing 1 - 15 )
Per Page 
Home  >  Flow, Level & Valves > Flowmeters > Flow Controllers
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title