United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$500 to $799
(2)
$800 to $1,099
(1)
Availability:
In Stock
(2)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Flow, Level & Valves
>
Level Control
>
Flowline Echotouch Transmitters and Controllers
> Flowline EchoSonic II Ultrasonic Level Transmitters
Flowline EchoSonic II Ultrasonic Level Transmitters
Optimize sensor placement with unmatched beam width for applications with restricted space
Industry-leading miniature dead band maximizes the filling capacity of vessels
Offered in 3 measurement ranges up to 32.8 ft (10 m)
WebCal, an innovative PC user interface provides fast and accurate configuration
NEMA 4X rated polycarbonate enclosure rated with PVDF transducer
The EchoSonic II
general purpose two-wire ultrasonic transmitter provides non-contact
level measurement up to 32.8 ft (10 m), and is well suited for challenging ultrapure, corrosive, slurry or waste liquids. The transmitter is broadly selected for atmospheric bulk storage, day tank, waste sump, process vessel, IBC tote, 55-gallon drum and waste sump applications. Media examples include acetic acid and resin.
The Fob USB interface tool (LI99-1001) is necessary to change EchoSonics from the factory configuration. One Fob will configure multiple EchoSonics.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
EchoSonic II 8 M range - 2" NPT threads w/Fob
Product Type Ultrasonic Level Controls,Material Housing: polypropylene
Transducer: PVDF,Brand Flowline
EW-23002-03
In stock.
$750.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Flowline EchoSonic II Ultrasonic Level Transmitters
EchoSonic II 3m range - 1
Product Type Ultrasonic Level Controls,Material Housing: polypropylene
Transducer: PVDF,Brand Flowline
EW-23002-02
In stock.
$550.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Flowline
EchoSonic II
Ultrasonic Level Transmitters
EchoSonic II 10.0M range - 2" NPT threads w/Fob
Product Type Ultrasonic Level Controls,Material Housing: polypropylene
Transducer: PVDF,Brand Flowline
EW-23002-04
Usually ships in 8 days.
$850.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Flowline EchoSonic II Ultrasonic Level Transmitters
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Flow, Level & Valves
>
Level Control
>
Flowline Echotouch Transmitters and Controllers
> Flowline EchoSonic II Ultrasonic Level Transmitters
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title