United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Pipettes and Pipettors
(996)
Microfluidics
(476)
Tanks and Drums
(196)
Syringes
(140)
Masterflex L/S Tubing Pumps
(94)
Burettes (Burets)
(66)
Centrifugal Separation
(60)
Titrators
(46)
Masterflex I/P Tubing Pumps
(41)
Piping
(26)
Masterflex C/L Tubing Pumps
(10)
Masterflex B/T Tubing Pumps
(7)
Brand:
Cole-Parmer
(222)
Masterflex
(150)
Thermo Scientific
(98)
[+]
Show more
(17)
Hamilton
(80)
Thermo Scientific Finnpipette
(76)
Eppendorf
(53)
C-FLEX
(39)
Ovation
(38)
Pyrex
(35)
B-D
(32)
Thermo Scientific Finntip
(32)
KIMBLE-CHASE
(31)
Kimax
(28)
ART
(28)
BrandTech
(24)
Finntip
(23)
SoftGrip
(16)
Thermo Scientific Heraeus
(14)
Micronit
(14)
Vistalabs
(12)
[-]
Show less
Product Type:
Pipette Tips
(162)
Adjustable Volume Pipettors
(139)
Capillary Column
(98)
[+]
Show more
(17)
Chromatography syringe
(89)
Fixed Volume Pipettors
(76)
Tips
(73)
Luer fittings
(71)
Accessory
(61)
Pump Drive
(61)
Peristaltic Pump
(58)
Tubing
(49)
Cylindrical Tanks
(46)
Multichannel Pipettors
(45)
Electronic Pipettors
(39)
Pipette Controllers
(38)
Syringe
(38)
Rectangular Tanks
(32)
Stainless Steel Tanks
(30)
Piping
(26)
Reservoirs
(20)
[-]
Show less
Price Range:
$0 to $100
(710)
$100 to $299
(578)
$300 to $499
(314)
[+]
Show more
(4)
$500 to $799
(171)
$800 to $1,099
(109)
$1,100 to $1,999
(113)
$2,000 and Over
(164)
[-]
Show less
Availability:
In Stock
(690)
Capacity (mL):
0 - 20
(43)
21 - 40
(17)
41 - 60
(20)
[+]
Show more
(4)
1.25
(3)
2.5
(6)
10 mL included; 20 and 50 mL optional
(1)
12.5
(1)
[-]
Show less
Material:
Glass
(83)
Nylon
(71)
HDPE
(45)
[+]
Show more
(9)
Polypropylene
(38)
Polyethylene
(35)
HDLPE
(26)
Polycarbonate
(22)
LLDPE
(17)
Stainless Steel (SS)
(16)
Stainless Steel
(14)
PTFE
(10)
Plastic
(10)
[-]
Show less
Autoclavable:
Yes
(56)
Control type:
Digital Variable Speed
(47)
Fixed speed
(24)
Analog Variable speed
(23)
[+]
Show more
(2)
Variable speed
(16)
Analog Variable Speed
(1)
[-]
Show less
Max volume (mL):
1
(47)
0.01
(43)
5
(42)
[+]
Show more
(17)
10
(34)
0.2
(29)
0.1
(28)
0.02
(22)
0.05
(19)
2
(17)
25
(15)
0.3
(13)
0.002
(12)
0.25
(9)
0.025
(9)
0.005
(6)
15
(5)
1.6
(1)
3.9
(1)
75
(1)
9
(1)
[-]
Show less
Max temperature:
150°F/65°C
(40)
220°F/104°C
(28)
140°F/60°C
(20)
[+]
Show more
(2)
650°F/182°C
(16)
500°F/260°C
(8)
[-]
Show less
Feet per pack:
1 to 5
(4)
20 to 50
(38)
100 and Above
(10)
CE Compliance:
Yes
(33)
Max RCF (x g):
25,314
(12)
17,000
(2)
Max rpm:
15,200
(12)
13,300
(1)
13300
(1)
Learn more:
Isolating Genomic DNA from Whole Blood
Tygon?« Flexible Plastic Tubing to Tubing Solvent Sealing
Bitter Brew
TanksResin Selection Guide
[+]
Show More
ISO 80369It?ÇÖs Coming to a Device Maker Near You
The Hidden Costs of Mercury-Filled Vacuum Gauges
Wine in Plastic Tanks? - A Case Study
InnoCal Pump Reconditioning Program
Thermo Scientific Finnpipette F1 antimicrobial feature
Equipping a Laboratory: A Study in Customer Service Excellence
PVC and CPVC Piping Selection Guide
Gulf Oil Spill Cleanup Puts Spotlight on Centrifugal Multipurpose Separators/Extractors
Physical Properties of Glass
Thermo Scientific Finntip?« Sterile and Finntip Filter Sterile - Importance of Guaranteed Purity
Masterflex?« C/L?« Low-Flow Tubing Pumps
Fittings Selection Guide
Basics of Cartridge Filtration
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Fluid & Liquid Handling
Fluid & Liquid Handling
Cole-Parmer is dedicated to providing the widest selection of quality pumps available. Whether you're involved in an experimental and research laboratory, the industrial marketplace or original equipment (OEM), we've got the right model for your application. Cole-Parmer offers many different pump technologies to meet your specific needs. These pump technologies include: air-operated double diaphragm (AODD), bellows, centrifugal - both sealless and mechanical sealed, diprhagm, dispensing, double-diaphragm, drum, flexible impeller, flexible liner, gear, hand, hose, Ismatec peristaltic, Masterflex peristaltic, peristaltic, piston, plunger, progressing cavity, roller, rotary lobe, rotary vane, submersible, and syringe to name a few. With the above mentioned pump tecnoligies we are able to provide pumps with flow rate capabilities from picoliter per minute flowranges to thousnds of gallon per minute to work well with your laboratory equipment and/or scientific equipment. Many www.coleparmer.com ads offer simple parts or consummables to provide a one stop shop for your replacement laboratory supplies. All to meet your laboratory, pilot plant of production facility needs.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Masterflex L/S Digital Drive, 600 RPM, 115/230 VAC
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.006
EW-07523-80
In stock.
$1,898.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex L/S Standard Digital Drives
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 6 to 600 rpm, 90 to 260 VAC.
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.36
EW-07528-10
In stock.
$1,120.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Precision Variable-Speed Console Drives
Masterflex L/S Digital Drive, 100 RPM, 115/230 VAC
Product Type Pump Drive,Max flow rate mL/min 566,Min flow rate mL/min 0.001
EW-07523-90
In stock.
$1,898.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Masterflex L/S Standard Digital Drives
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 1 to 100 rpm, 90 to 260 VAC.
Product Type Pump Drive,Max flow rate mL/min 580,Min flow rate mL/min 0.06
EW-07528-30
In stock.
$1,120.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Precision Variable-Speed Console Drives
Masterflex I/P Precision Brushless Drive, 33 to 650 rpm, 115/230 VAC
Product Type Masterflex I/P Drive,Max flow rate mL/min 19000,Min flow rate mL/min 200
EW-77410-10
In stock.
$2,035.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
I/P
Precision Brushless Drives
Masterflex L/S economy variable-speed drive, 20 to 600 rpm, 115 VAC.
Product Type Pump Drive,Max flow rate mL/min 2900,Min flow rate mL/min 1.2
EW-07554-90
In stock.
$638.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Economy Variable-Speed Console Drives
Masterflex L/S Digital Pump System with Easy-Load II Pump Head, 600 RPM, 115/230 VAC
Product Type Peristaltic Pump,Min flow rate mL/min 0.28,Max flow rate mL/min 1700
EW-77921-70
In stock.
$2,230.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S
Standard Digital Pump Systems
L/S computer-compatible/programmable digital drive, 0.1 to 600 rpm, 115/230 VAC
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.006
EW-07551-00
In stock.
$2,198.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Computer-Compatible Digital Drives
Clear C-FLEX Tubing, 1/2"ID x 3/4"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-18
In stock.
$137.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Clear C-FLEX Tubing
Masterflex L/S Precision Modular Drive with Bench Top Controller and Remote I/O; 6 to 600 rpm, 90 to 260 VAC
Product Type Pump Drive,Max flow rate mL/min 3400,Min flow rate mL/min 0.36
EW-07557-00
In stock.
$1,190.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Transfer Pipette 7.5ml Graduated Large Bulb 500/pk
Product Type Transfer Pipettes,Max volume mL 7.5,Max volume µL 7500
EW-06226-63
In stock.
$45.00 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Disposable Transfer Pipettes
Clear C-FLEX Tubing, 3/8"ID x 5/8"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-16
In stock.
$97.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Clear C-FLEX Tubing
Masterflex L/S economy variable-speed drive, 7 to 200 rpm, 115 VAC.
Product Type Pump Drive,Max flow rate mL/min 1130,Min flow rate mL/min 0.42
EW-07554-80
In stock.
$638.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
L/S Economy Variable-Speed Console Drives
Clear C-FLEX Tubing, 3/4 x 1-1/8", 15 ft/pk
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-21
In stock.
$269.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Clear C-FLEX Tubing
Masterflex I/P Precision Brushless Drive with Analog Remote, 33 to 650 rpm, 115/230 VAC
Product Type Masterflex I/P Drive,Max flow rate mL/min 26000,Min flow rate mL/min 200
EW-77411-00
In stock.
$2,620.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
I/P
Precision Brushless Drives with Remote Capabilities
2158
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
Fluid & Liquid Handling
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title