Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 

Formaldehyde

Formaldehyde
Choose from our comprehensive line of Formaldehyde and other FMOCC to Formyl products.
 Product Detail
Sort
 Availability
 Price

Thermo Scientific Richard-Allan Scientific Formaldehyde Solution 37%, 4 x 1 Gal

  • Product Type: Inorganic Chemical
  • Brand: Thermo Scientific
  • Manufacturer number: 9311
EW-48800-00
Usually ships in 13 days.
$134.00 USD/CASE

Qty:

Cole-Parmer Formaldehyde, 37% w/w, ACS Grade, 500 mL

  • Product Type: Inorganic Chemical
  • Purity grade: ACS
  • CAS Number: 7732-18-5, 50-00-0, 67-56-1
EW-88405-59
Available for order
or call 1-888-358-4705 to order
$34.00 USD/EACH

Qty:

Formaldehyde (37% by Weight/Histological) (1 gal.)

  • Product Type: Organic Chemicals
  • Purity grade: Histological
  • CAS Number: 50000
EW-88010-20
Available for order
or call 1-888-358-4705 to order
$228.00 USD/EACH

Qty:

Cole-Parmer Formaldehyde, 37% w/w, ACS Grade, 1 L

  • Product Type: Inorganic Chemical
  • Purity grade: ACS
  • CAS Number: 7732-18-5, 50-00-0, 67-56-1
EW-88405-57
Available for order
or call 1-888-358-4705 to order
$53.00 USD/EACH

Qty:

Formaldehyde, 37 wt% solution in water, stabilized with 10-15% methanol (1l)

  • Product Type: Chemical Reagents
  • CAS Number: 50-00-0
  • Form: Liquid
EW-88116-10
Usually ships in 5 days.
$46.50 USD/EACH

Qty:

Cole-Parmer Formaldehyde, 37% w/w, ACS Grade, 20 L

  • Product Type: Inorganic Chemical
  • Purity grade: ACS
  • CAS Number: 7732-18-5, 50-00-0, 67-56-1
EW-88405-58
Available for order
or call 1-888-358-4705 to order
$202.00 USD/EACH

Qty:

Formaldehyde, 37 wt% solution in water, stabilized with 10-15% methanol (2.5l)

  • Product Type: Organic Chemicals
  • CAS Number: 50-00-0 and 7732-18-5 and 67-56-1
  • Form: Liquid
EW-88116-11
Usually ships in 2 days.
$60.50 USD/EACH

Qty:

Formaldehyde (37% by Weight/Molecular Biology) (500mL)

  • Product Type: Chemical Bio-Reagents
  • Purity grade: Molecular Biology
  • CAS Number: 50-00-0 (Formaldehyde), 67-56-1 (Methyl Alcohol)
EW-88067-65
Usually ships in 3 days.
$83.00 USD/EACH

Qty:

Formaldehyde, 37 wt% solution in water, stabilized with 10-15% methanol (5l)

  • Product Type: Organic Chemicals
  • CAS Number: 50-00-0
  • Form: Liquid
EW-88116-12
Usually ships in 4 days.
$79.25 USD/EACH

Qty:

Formaldehyde, reagent ACS, 37 wt% solution, stabilized (100g)

  • Product Type: Chemical Reagents
  • CAS Number: 50-00-0
  • Form: Liquid
EW-88347-71
Usually ships in 2 days.
$49.00 USD/EACH

Qty:

Formaldehyde (37% by Weight. With Preservative./Certified ACS) (1 liter)

  • Product Type: Organic Chemicals
  • Purity grade: ACS Reagent Grade
  • CAS Number: 50000
EW-88010-16
Usually ships in 2 days.
$165.00 USD/EACH

Qty:

Formaldehyde (37% by Weight/Histological) (20 liter)

  • Product Type: Chemical Reagents
  • Purity grade: Histological
  • CAS Number: 50-00-0
EW-88073-37
Usually ships in 5 days.
$430.00 USD/EACH

Qty:

Formaldehyde (40% by Volume/USP) (4 liter)

  • Product Type: Organic Chemicals
  • Purity grade: USP
  • CAS Number: 50-00-0
EW-88073-41
Usually ships in 2 days.
$237.00 USD/EACH

Qty:

Formaldehyde (37% by Weight/Molecular Biology) (25mL)

  • Product Type: Chemical Bio-Reagents
  • Purity grade: Molecular Biology
  • CAS Number: 50-00-0 (Formaldehyde), 67-56-1 (Methyl Alcohol)
EW-88067-64
Usually ships in 2 days.
$61.00 USD/EACH

Qty:

Formaldehyde, reagent ACS, 37 wt% solution, stabilized (1kg)

  • Product Type: Chemical Reagents
  • CAS Number: 50-00-0
  • Form: Liquid
EW-88347-70
Usually ships in 2 days.
$115.00 USD/EACH

Qty:

19 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#