log in or register
2,600 pages
of answers
FREE
2013/14 Cole-Parmer
Sourcebook
Our General Catalogue features 60,000+ products for electrochemistry, fluid handling, industrial process, laboratory research, facility and personal safety, and more.
Request your copy today.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Freeze Dryers
Freeze Dryers
Cole-Parmer offers a range of benchtop freeze dryers that are available with system capacities from 1L, 2.5L, 4.5L and 6.5L. Upon request with our technical staff we are able to offer console sizes up to 18L in capacities. Our range in freeze dryers include variations in refrigerator collectors that cool to -50°C and -84°C as well as PTFE-coated models for more corrosive compounds. Our freeze dry systems are designed to handle the lyophilization needs of research and pilot plant laboratories, with the lower temperature collectors ideal for freeze drying samples with low eutectic temperature.
Freeze drying involves the removal of water or other solvents from a frozen product. Freeze dryers carry out three steps for this process - prefreezing, primary drying and finally secondary drying. Freeze drying has been used in a number of applications however it is most commonly used in the pharmaceutical industry, for example stabilization of living material such as microbial cultures, preservation of animal specimens and the concentration and recovery of reaction products. Once freeze dried properly, products do not need refrigeration and items can be stored at ambient temperatures.
To complete your freeze drying systems, you will require the following components:
  • Freeze Dryer - Cole-Parmer's catalog range carries capacities up to 6L with temperatures of -54°C and -84°C, however larger capacities of up to 18L are available upon request.
  • Drying System Platforms - Depending on your application, Cole-Parmer's freeze dryers offer a range of drying platforms. The drying platforms are the interfacing device that conducts solvent from the sample to the freeze dryer and may connect or hold sample containers.
  • Chamber platform
    Either accommodates sample shelving or provides flexible port connectivity for up to 12 or 16 sample flasks. Shelves maybe used for either bulk samples or bottles and are available with heaters to accelerate the freeze drying process.
    Manifold
    For parallel lyophilization of small sample volumes and products with high eutectic and collapse temperatures. Sample is prefrozen and then attached to the manifold with vacuum so that the product is not warmed. This process allows for samples of different makeup and size to be attached to the system, and for the sample to be removed from the manifold as the product is dried.
    Stoppering Tray Dryer
    For batches of small vessels or bulk sample on shelves. Ideal for preparing a large quantity of vials containing the same sample. The samples can be prefrozen directly on the shelf of the tray dryer. This method ensures that all samples are handled alike, also allowing sample vials to be vacuum sealed in situ at the same time and under the same conditions.
  • Vacuum Pump - Providing an environment that allows for easy removal of water molecules from the sample is critical to successful freeze drying. A vacuum pump is needed to lower the pressure around the product being freezed. Select from standard or chemical resistant models to enable freeze drying with more aggressive solvents
  • Sample containers - Sturdy, borosilicate glass vessels options include flasks, serum bottles, vials, and ampules
  • Add this option:
    • Shell Freezer - for preliminary freezing of samples. Improves freeze drying efficiency by maximizing sample surface area in cylindrical fast free flasks. Available as stand-alone or integrated unit.
    Typical Application for freeze dryers
    • Freeze Drying Fish Samples for Volatile Substances
    • General Freeze Dry Application for Snake Venom
    • Freeze Drying Fungi
    • General Freeze Dry Application for Monitoring Soil Samples
    • Freeze Dry Applications for Monitoring Anti-Cancer Drugs
    • Freeze Drying Mango Powder
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
     Product Detail
    Sort
     Availability
     Price
    Representative photo only 2 5 L Benchtop Cascade Freeze Dry System 84 with SS interior 220V
    2.5 L Benchtop Cascade Freeze Dry System, -84° with SS interior, 220V
    Product Type Freeze Dryer,Capacity 2.5 L,Capacity Liters 2.5
    EW-03339-20
    Call 1-888-358-4705 for availability.
    $11,520.00 USD/EACH
    Qty:
    Representative photo only 1 L Benchtop Freeze Dry System 54 with SS interior 220V
    1 L Benchtop Freeze Dry System, -54° with SS interior, 220V
    Product Type Freeze Dryer,Capacity 1 L,Capacity Liters 1
    EW-03337-10
    Call 1-888-358-4705 for availability.
    $5,965.00 USD/EACH
    Qty:
    EW-03337-00 1 L Benchtop Freeze Dry System, -54° with SS interior, 115V
    1 L Benchtop Freeze Dry System, -54 with SS interior, 115V
    Product Type Freeze Dryer,Capacity 1 L,Capacity Liters 1
    EW-03337-00
    Call 1-888-358-4705 for availability.
    $5,870.00 USD/EACH
    Qty:
    Representative photo only Drying chamber with 12 ports
    Drying chamber with 12 ports
    Product Type Drying Chambers,Capacity Ports:12
    EW-03346-01
    Call 1-888-358-4705 for availability.
    $1,790.00 USD/EACH
    Qty:
    See all Drying Chambers
    EW-03336-97 Labconco<small><sup>&reg;</sup></small> Stoppering Tray Dryer, 115V/60Hz
    Labconco Stoppering Tray Dryer, 115V/60Hz
    Product Type Stoppering Tray Dryers,Capacity 3 shelves, 196 sq.in.,Power VAC 115
    EW-03336-97
    Call 1-888-358-4705 for availability.
    $23,535.00 USD/EACH
    Qty:
    Representative photo only 2 5 L Benchtop Freeze Dry System 54 with PTFE interior 115V
    2.5 L Benchtop Freeze Dry System, -54° with PTFE interior, 115V
    Product Type Freeze Dryer,Capacity 2.5 L,Capacity Liters 2.5
    EW-03338-05
    Call 1-888-358-4705 for availability.
    $7,875.00 USD/EACH
    Qty:
    EW-03337-66 8 Port Clear Drying Chambers
    8 Port Clear Drying Chambers
    Product Type Drying Chambers
    EW-03337-66
    Call 1-888-358-4705 for availability.
    $1,990.00 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 300 mL
    Fast-Freeze flask, 300 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-04
    Usually ships in 14 days.
    $197.00 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 40 mL
    Fast-Freeze flask, 40 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-00
    Usually ships in 14 days.
    $103.00 USD/EACH
    Qty:
    Representative photo only Fast Freezeflask 1200 mL
    Fast-Freezeflask, 1200 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-07
    Usually ships in 14 days.
    $296.00 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 150 mL
    Fast-Freeze flask, 150 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-03
    Usually ships in 14 days.
    $167.00 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 80 mL
    Fast-Freeze flask, 80 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-01
    Usually ships in 14 days.
    $91.75 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 2000 mL
    Fast-Freeze flask, 2000 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-08
    In stock.
    $350.00 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 600 mL
    Fast-Freeze flask, 600 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-05
    Usually ships in 14 days.
    $245.00 USD/EACH
    Qty:
    Representative photo only Fast Freeze flask 120 mL
    Fast-Freeze flask, 120 mL
    Material Glass,Brand Fast-Freeze
    EW-03353-02
    Call 1-888-358-4705 for availability.
    $161.00 USD/EACH
    Qty:
    65 results (showing 1 - 15 )
    Per Page 
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title