Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Cooling Equipment > Freezers

Freezers

Freezers

Buying a laboratory freezer isn’t like picking up an appliance for your new home. There are few other factors to consider when purchasing a laboratory freezer either for a sterile or controlled environment. Consider an upright freezer which cools your samples down to -86ºC. While an undercounter laboratory freezer allows you to save space in even the most crowded lab. A general-purpose laboratory freezer is ideal for general storage of samples or materials. For more critical samples, choose an ultra-low temperature laboratory freezer. Cole- Parmer features the following brands: Thermo Scientific REVCO, Nor-Lake Scientific, and ScienTemp. Don’t forget your laboratory freezer accessories including storage racks and boxes, chart recorders, remote alarm systems, and more. If you have questions about which laboratory freezer to choose for your workspace contact a Cole-Parmer TechnicalExpert today.
 Product Detail
Sort
 Availability
 Price
Representative photo only Thermo Scientific Revco DxF Series 40C Upright Freezer 23 cubic feet 400box 115V 60Hz

Thermo Scientific Revco DxF Series, -40°C Upright Freezer, 23 cubic feet (400box), 115V/60Hz

Product Type Ultra-Low Freezer (upright),Capacity cu ft 23,Capacity Liters 653
EW-44146-37
Usually ships in 13 days.
$12,970.00 USD/EACH

Qty:

EW-44204-00 Compact ultra-low freezer, benchtop, 115 VAC, 60 Hz

Compact ultra-low freezer, benchtop, 115 VAC, 60 Hz

Product Type Benchtop Freezers,Capacity cu ft 1,Capacity Liters 28
EW-44204-00
Call 1-888-358-4705 for availability.
$4,395.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Revco ExF Series 86C Upright Freezer 17 3 cubic feet 320box 208 230V 60Hz

Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 17.3 cubic feet (320box), 208-230V/60Hz

Product Type Ultra-Low Freezer (upright),Capacity cu ft 17.3,Capacity Liters 490
EW-44146-24
Usually ships in 13 days.
$14,730.00 USD/EACH

Qty:

Representative photo only Standard low temperature cabinets 24 9 cu ft 34C 115 VAC

Standard low-temperature cabinets; 24.9 cu ft; -34°C; 115 VAC

Product Type Freezer (chest),Capacity cu ft 24.9,Capacity Liters 705
EW-03815-36
Call 1-888-358-4705 for availability.
$2,795.00 USD/EACH

Qty:

Representative photo only Space save low temperature cabinets 3 1 cu ft 85C 115 VAC

Space-save low-temperature cabinets; 3.1 cu ft; –85°C; 115 VAC

Product Type Freezer (chest),Capacity cu ft 3.1,Capacity Liters 88
EW-03815-40
Call 1-888-358-4705 for availability.
$5,210.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Revco DxF Series 40C Upright Freezer 17 3 cubic feet 320box 208 230V 60Hz

Thermo Scientific Revco DxF Series, -40°C Upright Freezer, 17.3 cubic feet (320box), 208-230V/60Hz

Product Type Ultra-Low Freezer (upright),Capacity cu ft 17.3,Capacity Liters 490
EW-44146-35
Usually ships in 13 days.
$10,990.00 USD/EACH

Qty:

Representative photo only Space save low temperature cabinets 3 1 cu ft 45C 115 VAC

Space-save low-temperature cabinets; 3.1 cu ft; –45°C; 115 VAC

Product Type Freezer (chest),Capacity cu ft 3.1,Capacity Liters 88
EW-03815-38
Call 1-888-358-4705 for availability.
$3,635.00 USD/EACH

Qty:

Representative photo only Freezer LED Display with Alarm 24 cu ft with Casters 115V 60Hz

Freezer, LED Display with Alarm, 24 cu ft with Casters, 115V/60Hz

Product Type Freezer (upright),Capacity cu ft 24,Capacity Liters 680
EW-44173-64
Usually ships in 3 days.
$4,723.00 USD/EACH

Qty:

Representative photo only Shuttle Portable Ultra Low Temperature Freezer 120 220V US std Plug

Shuttle Portable Ultra-Low Temperature Freezer, 120/220V US std. Plug

Product Type Portable Ultra-Low Temperature Freezer,Capacity cu ft 0.9,Capacity Liters 25
EW-44199-05
Usually ships in 4 days.
$5,799.00 USD/EACH

Qty:

Representative photo only Chest style freezer 20 3 cu ft 34C 115V

Chest-style freezer, 20.3 cu ft, -34°C, 115V

Product Type Freezer (chest),Capacity cu ft 20.3,Capacity Liters 566
EW-03815-47
Usually ships in 15 days.
$2,525.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Revco Lab freezer 20C 115 VAC 60 Hz

Thermo Scientific Revco Lab freezer, -20°C, 115 VAC, 60 Hz

Product Type Enzyme Freezers,Capacity cu ft 23.3,Capacity Liters 659
EW-44164-10
Usually ships in 4 days.
$7,030.00 USD/EACH

Qty:

EW-44261-50 StableTemp freezer, single solid door model

StableTemp 24 cu ft freezer stainless steel exterior w/alarms, access port, solid door and casters; 115V

Product Type General Purpose Freezer,Capacity cu ft 24,Capacity Liters 679
EW-44261-50
Usually ships in 13 days.
$7,680.00 USD/EACH

Qty:

EW-44147-14 Thermo Scientific Revco<small><sup>&reg;</sup></small> CxF Series, -86&deg;C Chest Freezer is shown here for illustrative purposes only. Please refer to product description.

Thermo Scientific Revco CxF -86C Chest Freezer, 12.7 cubic feet , 115V/60Hz

Product Type Chest Freezer,Capacity cu ft 12.7,Capacity Liters 360
EW-44147-14
Usually ships in 13 days.
$12,010.00 USD/EACH

Qty:

Representative photo only Standard low temperature cabinets 6 8 cu ft 45C 115V

Standard low-temperature cabinets; 6.8 cu ft; -45°C; 115V

Product Type Freezer (chest),Capacity cu ft 6.8,Capacity Liters 193
EW-03815-23
Call 1-888-358-4705 for availability.
$3,915.00 USD/EACH

Qty:

EW-03815-22 Standard low-temperature cabinets

Standard low-temperature cabinets; 9.4 cu ft; -40C; 115V

Product Type Freezer (chest),Capacity cu ft 9.4,Capacity Liters 266
EW-03815-22
Call 1-888-358-4705 for availability.
$4,050.00 USD/EACH

Qty:

151 results (showing 1 - 15 )
Per Page 
Home  >  Cooling Equipment > Freezers

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title