United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Cooling Equipment
>
Ice Machines
> Frigimat Junior Dry Ice Maker
Frigimat Junior Dry Ice Maker
Produces dry ice in flake or snow
Produces only the dry ice you need
Quick and easy to use
Operate these dry ice makers by screwing the nozzle directly to cylinder of compressed LCO
2
.
Note:
cylinder must have a dip tube. Bag is secured to cone on the nozzle with the clamp. Compressed LCO
2
expands into the bag and creates dry-ice snow. The ice may be squeezed into a hard pack or left in flake form depending on intended use. It makes 0.77 kg (1.7 lb) of flakes per bag. The cone is 13 cm (5") long x 6 cm (2-5⁄16") diameter at end. The bag is 15 cm (6") W x 30 cm (12") L.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Junior dry ice maker
Product Type Dry Ice Makers,Description Junior Dry Ice Maker,Brand Frigimat
EW-03716-32
In stock.
$170.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Frigimat Junior Dry Ice Maker
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Cooling Equipment
>
Ice Machines
> Frigimat Junior Dry Ice Maker
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title