log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Furniture & Fume Hoods > Fume Extractors
Fume Extractors
Fume Extractors
Cole-Parmer's fume extractors provide localized ventilation at the contaminant source. Most fume extractors have extractor arms that can be easily positioned to remove fumes or air-borne particulates. These air cleaning systems capture and remove smoke, dust, mist, and fumes before they enter the work area, then recirculate clean air back into the workplace.
Two-arm extractor units are Ideal for two-station setups. You can use an included inlet cap to convert to single-arm use. Single arm use will increase the air volume which will provide more exhaust power at a single station. Fume extractors are often used when the fume source can not be readily moved to a conventional hood or when fumes are innocuous, bad-smelling without being chemically dangerous. Cole-Parmer offers stationary, portable, bench top, and downdraft stations that are suitable for many applications, including welding and soldering stations, metal grinding, solvent mixing, and even painting. Fume extractors use high efficiency HEPA and carbon-activated filters that are replaceable to ensure only clean air is vented back into the surrounding environment.
 Product Detail
Sort
 Availability
 Price
Representative photo only Compact Ductless Air Cleaner 350W Black
Compact Ductless Air Cleaner, 350W, Black
Product Type Fume hood extractor systems,Exterior Dimensions " W 12.5,Exterior Dimensions " H 12.5
EW-33703-39
Usually ships in 10 days.
$1,324.00 USD/EACH
Qty:
EW-33703-25 Shown with filter (order seperately)
Benchtop air cleaning system
Product Type Fume extractor systems,Exterior Dimensions " W 12,Exterior Dimensions " H 10
EW-33703-25
Usually ships in 5 days.
$1,295.00 USD/EACH
Qty:
Representative photo only Extractor Arm 7
Extractor Arm, 7'
Product Type Fume hood extractor systems
EW-33700-01
Usually ships in 14 days.
$1,409.00 USD/EACH
Qty:
EW-33703-30 Portable air cleaning system
Portable air cleaning system
Product Type Fume extractor systems,Exterior Dimensions " W 20.5,Exterior Dimensions " H 36.8
EW-33703-30
Usually ships in 8 days.
$1,846.00 USD/EACH
Qty:
Representative photo only 1 hp Fan for 33700 series 115 230V
1-hp Fan for 33700 series, 115/230V
Product Type Fume hood extractor systems,Blower hp 1,Power VAC 115/230
EW-33700-60
Usually ships in 14 days.
$1,217.00 USD/EACH
Qty:
Representative photo only 3 4 hp Fan for 33700 series 115 230V
3/4-hp Fan for 33700 series, 115/230V
Product Type Fume hood extractor systems,Blower hp 1/2,Power VAC 115/230
EW-33700-50
Usually ships in 13 days.
$988.00 USD/EACH
Qty:
EW-33704-90 SECONDARY MERV 8 PLEATED FILTER
Secondary Merv 8 Pleated Filter
Brand AIR IMPURITIES REMOVAL SYSTEM
EW-33704-90
Usually ships in 8 days.
$56.00 USD/EACH
Qty:
See all
Representative photo only Extractor Arm 14 ft
Extractor Arm, 14-ft
Product Type Fume hood extractor systems
EW-33700-04
Usually ships in 14 days.
$2,003.00 USD/EACH
Qty:
Representative photo only Extractor Arm 10 ft
Extractor Arm, 10-ft
Product Type Fume hood extractor systems
EW-33700-02
Usually ships in 14 days.
$1,641.00 USD/EACH
Qty:
EW-33704-02 EXTRACT-ALL<TM> PORTABLE AIR CLEANING SYSTEM WITH 4&APOS; TO 6&APOS; LONG EXTRACTOR ARM, 120 VAC
EW-33704-02
Usually ships in 8 days.
$3,942.00 USD/EACH
Qty:
See all
Representative photo only Extractor Arm Light 14 ft
Extractor Arm & Light, 14-ft
Product Type Fume hood extractor systems,Power VAC 115
EW-33700-14
Usually ships in 14 days.
$2,599.00 USD/EACH
Qty:
Representative photo only Wall Bracket for 33700 Series Extractor Arms
Wall Bracket for 33700 Series Extractor Arms
Product Type Fume hood extractor systems
EW-33700-90
Usually ships in 7 days.
$94.00 USD/EACH
Qty:
EW-33704-91 PRIMARY MERV 14 CELL FILTER
Primary Merv 14 Cell Filter
Brand AIR IMPURITIES REMOVAL SYSTEM
EW-33704-91
Usually ships in 8 days.
$186.00 USD/EACH
Qty:
See all
Representative photo only Extractor Arm Light 10 ft
Extractor Arm & Light, 10-ft
Product Type Fume hood extractor systems,Power VAC 115
EW-33700-12
Usually ships in 14 days.
$2,608.00 USD/EACH
Qty:
Representative photo only Extractor Arm Light 7 ft
Extractor Arm & Light, 7-ft
Product Type Fume hood extractor systems,Power VAC 115
EW-33700-10
Usually ships in 14 days.
$2,387.00 USD/EACH
Qty:
20 results (showing 1 - 15 )
Per Page 
Home  >  Furniture & Fume Hoods > Fume Extractors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title