log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  pH Meters and Water Quality Products > Electrodes, Industrial > Conductivity Electrodes - Industrial > +GF+ Signet Conductivity/Resistivity Electrodes with Integral Electronics
+GF+ Signet Conductivity/Resistivity Electrodes with Integral Electronics
+GF+ Signet Conductivity/Resistivity Electrodes with Integral Electronics
  • Integral mount systems for quick and easy installation
  • Compact design for maximum installation flexibility
  • Digital (S3L) interface or two-wire 4 to 20 mA output
  • EasyCal with automatic test solution recognition
+GF+ Signet Conductivity/Resistivity Sensors with Integral Electronics are available in various configurations for maximum installation flexibility. These combined integral system configurations allow for in-line mounting and include conductivity electrodes in a choice of K = 0.01, 0.1, 1.0, or 10.0 cell constants. Users have a choice of two outputs: digital (S3L) or 4 to 20 mA. The digital (S3L) output version allows for up to six sensor inputs directly into the Signet 8900 Multi-Parameter Controller. The two-wire 4 to 20 mA output is available with eight 4 to 20 mA output ranges for each electrode cell constant. Additionally, each range can be inverted and are field selectable by the user.
All units are built with NEMA 4X/IP 65 enclosures which allow wiring connections with long cable runs of up to 1,000 feet (305 m). EasyCal is a standard feature that automatically recognizes conductivity test solution values for simple field calibration.
 Product Detail
Sort
 Availability
 Price
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 1 0 4 to 20 mA output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 1.0, 4 to 20 mA output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-06
Usually ships in 10 days.
$804.00 USD/EACH
Qty:
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 0 1 4 to 20 mA output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 0.1, 4 to 20 mA output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-56560-33
Usually ships in 5 days.
$804.00 USD/EACH
Qty:
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 0 01 4 to 20 mA output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 0.01, 4 to 20 mA output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-05
Usually ships in 10 days.
$804.00 USD/EACH
Qty:
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 10 0 Digital S3L output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 10.0, Digital S3L output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-04
Usually ships in 10 days.
$785.00 USD/EACH
Qty:
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 1 0 Digital S3L output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 1.0, Digital S3L output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-03
Usually ships in 10 days.
$664.00 USD/EACH
Qty:
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 0 1 Digital S3L output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 0.1, Digital S3L output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-01
Usually ships in 10 days.
$664.00 USD/EACH
Qty:
Representative photo only  GF Signet Conductivity Cell with Integral Electronics K 10 0 4 to 20 mA output 3 4 NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 10.0, 4 to 20 mA output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-56560-34
Usually ships in 7 days.
$925.00 USD/EACH
Qty:
EW-19007-02 +GF+ Signet Conductivity Cell with Integral Electronics, K = 0.01, Digital S3L output, 3/4" NPT threads
+GF+ Signet Conductivity Cell with Integral Electronics, K = 0.01, Digital S3L output, 3/4" NPT threads
Product Type Conductivity Cell,Max temperature °C 85,Min temperature °C 10
EW-19007-02
Usually ships in 10 days.
$664.00 USD/EACH
Qty:
8 results (showing 1 - 8 )
Per Page 
Home  >  pH Meters and Water Quality Products > Electrodes, Industrial > Conductivity Electrodes - Industrial > +GF+ Signet Conductivity/Resistivity Electrodes with Integral Electronics
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title