log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Gas Detectors > Single Gas Detection > GasAlertClip Extreme Personal Monitors
GasAlertClip Extreme Personal Monitors
GasAlertClip Extreme Personal Monitors
  • Compact and easy to use—no maintenance required!
Operated by an advanced microcontroller and electrochemical sensor, the GasAlertClip Extreme monitors provide continuous, maintenance-free operation for up to two years from the day of activation. These disposable battery-operated monitors are easy to use—simply clip the meter to your pocket or belt and go!
View important gas-reading information and detector life remaining data on the full-feature LCD display. Monitors feature audible, visible, and vibrating alarms. Each monitor is encased in a shock- and water-resistant ABS enclosure and offers RFI protection to withstand the rigors of industrial and field use.
 Product Detail
Sort
 Availability
 Price
Representative photo only GasAlertClip Extreme Personal Monitor Oxygen 2 year operational life
GasAlertClip Extreme Personal Monitor, Oxygen, 2-year operational life
Product Type Personal Gas Monitors/Detectors,CE Compliance Yes,Brand GasAlertClip
EW-86383-05
Usually ships in 7 days.
$189.00 USD/EACH
Qty:
Representative photo only GasAlertClip Extreme Personal Monitor Sulfur Dioxide 2 year operational life
GasAlertClip Extreme Personal Monitor, Sulfur Dioxide, 2-year operational life
Product Type Personal Gas Monitors/Detectors,CE Compliance Yes,Brand GasAlertClip
EW-86383-07
Usually ships in 7 days.
$310.00 USD/EACH
Qty:
Representative photo only GasAlertClip Extreme Personal Monitor Hydrogen sulfide 3 year operational life
GasAlertClip Extreme Personal Monitor, Hydrogen sulfide, 3-year operational life
Product Type Personal Gas Monitors/Detectors,CE Compliance Yes,Brand GasAlertClip
EW-86383-09
Usually ships in 7 days.
$240.00 USD/EACH
Qty:
Representative photo only GasAlertClip Extreme Personal Monitor Carbon monoxide 2 year operational life
GasAlertClip Extreme Personal Monitor, Carbon monoxide, 2-year operational life
Product Type Personal Gas Monitors/Detectors,CE Compliance Yes,Brand GasAlertClip
EW-86383-01
Usually ships in 7 days.
$189.00 USD/EACH
Qty:
Representative photo only GasAlertClip Extreme Personal Monitor Carbon monoxide 3 year operational life
GasAlertClip Extreme Personal Monitor, Carbon monoxide, 3-year operational life
Product Type Personal Gas Monitors/Detectors,CE Compliance Yes,Brand GasAlertClip
EW-86383-11
Usually ships in 7 days.
$240.00 USD/EACH
Qty:
EW-86383-03 GasAlertClip<small><sup>&#153;</sup></small> Extreme Personal Monitor, Hydrogen sulfide, 2-year operational life
GasAlertClip Extreme Personal Monitor, Hydrogen sulfide, 2-year operational life
Product Type Personal Gas Monitors/Detectors,CE Compliance Yes,Brand GasAlertClip
EW-86383-03
Usually ships in 7 days.
$189.00 USD/EACH
Qty:
6 results (showing 1 - 6 )
Per Page 
Home  >  Safety Products > Gas Detectors > Single Gas Detection > GasAlertClip Extreme Personal Monitors
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title