log in or register
Product Type:
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
GasAlertQuattro Multigas Monitors
GasAlertQuattro Multigas Monitors
  • IntelliFlash green LED verifies operation and compliance to user and supervisor
  • Bright, extended visual alarm bars for viewing at any angle
Compact and lightweight design makes these gas detectors comfortable to wear all day. Continuous LCD displays real-time gas concentrations. Built-in concussion-proof boot and IP66/67 rating makes these detectors suitable for field use. Verify operation from a distance by looking for the IntelliFlash green LED with user-defined flash frequency. Additional features include one-button operation, data/event logging, audible/visual/vibrating alarms, user adjustable alarm set points, sensor self-test, and multilanguage capability (English, French, German, Spanish, and Portuguese). Approvals: CE; CSA Class I, Div. 1, Groups A, B, C, D; ATEX
 Product Detail
Sort
 Availability
 Price
EW-86418-60 GasAlertQuattro 4-Gas Detector, alkaline, %LEL, O2, H2S, CO
GasAlertQuattro 4-Gas Detector, alkaline, %LEL, O2, H2S, CO
Product Type 4 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-60
Usually ships in 7 days or sooner.
$695.00 USD/EACH
Qty:
Representative photo only GasAlertQuattro 4 Gas Detector Rechargeable LEL O2 H2S CO
GasAlertQuattro 4-Gas Detector, Rechargeable, %LEL, O2, H2S, CO
Product Type 4 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, 95dB audible alarm
EW-86418-62
Usually ships in 7 days or sooner.
$755.00 USD/EACH
Qty:
EW-86418-70 Model 86418-60 shown
GasAlertQuattro 3-Gas Detector, rechargeable, %LEL, O2, CO
Product Type 3 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-70
Usually ships in 7 days.
$705.00 USD/EACH
Qty:
EW-86418-72 Model 86418-60 shown
GasAlertQuattro 3-Gas Detector, alkaline, O2, H2S, CO
Product Type 3 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-72
Usually ships in 7 days.
$645.00 USD/EACH
Qty:
EW-86418-98 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, Rechargeable, %LEL, H2S
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-98
Usually ships in 7 days.
$655.00 USD/EACH
Qty:
EW-86418-78 Model 86418-60 shown
GasAlertQuattro 3-Gas Detector, Rechargeable, %LEL, H2S, CO
Product Type 3 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-78
Usually ships in 7 days.
$705.00 USD/EACH
Qty:
EW-86418-74 Model 86418-60 shown
GasAlertQuattro 3-Gas Detector, rechargeable, O2, H2S, CO
Product Type 3 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-74
Usually ships in 7 days.
$705.00 USD/EACH
Qty:
EW-86418-94 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, Rechargeable, O2, CO
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-94
Usually ships in 7 days.
$655.00 USD/EACH
Qty:
EW-86418-68 Model 86418-60 shown
GasAlertQuattro 3-Gas Detector, alkaline, %LEL, O2, CO
Product Type 3 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-68
Usually ships in 7 days.
$645.00 USD/EACH
Qty:
EW-86418-82 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, rechargeable, %LEL, O2
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-82
Usually ships in 7 days.
$655.00 USD/EACH
Qty:
EW-86418-88 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, Alkaline, O2, H2S
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-88
Usually ships in 7 days.
$595.00 USD/EACH
Qty:
EW-86418-66 Model 86418-60 shown
GasAlertQuattro 3-Gas Detector, rechargeable, %LEL, O2, H2S
Product Type 3 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-66
Usually ships in 7 days.
$710.00 USD/EACH
Qty:
EW-86419-04 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, Alkaline, %LEL, CO
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86419-04
Usually ships in 7 days.
$595.00 USD/EACH
Qty:
EW-86418-80 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, Alkaline, %LEL, O2
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-80
Usually ships in 7 days.
$595.00 USD/EACH
Qty:
EW-86418-90 Model 86418-60 shown
GasAlertQuattro 2-Gas Detector, Rechargeable, O2, H2S
Product Type 2 Gas Monitors/Detectors,CE Compliance Yes,Alarm six red LEDs visual alarm, vibrating alarm, and 95dB audible alarm
EW-86418-90
Usually ships in 7 days.
$655.00 USD/EACH
Qty:
22 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title