Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Product Type:
Availability:
In Stock (107)
CE Compliance:
Yes (28)
No. of wells:
10 (25)
14 (20)
12 (10)
20 (8)
27 (6)
16 (5)
15 (3)
8/10 (3)
12/16 (3)
10/14 (3)
8/16 (3)
13 (3)
26 (3)
28 (3)
51 (3)
102 (3)
4 (2)
Buffer solution required (mL):
1600 (16)
250 (3)
300 (3)
2300 (2)
700 (2)
400 (1)
1000 (1)
4500 (1)
1400 (1)
3000 (1)
150 (1)
750 (1)
1500 (1)
725 (1)
500 (1)
825 (1)
900 (1)
50 mL (1)
Gel thickness (mm):
1.5 (15)
Gel size (cm L):
8 (10)
25 (4)
10 (3)
40 (1)
8.5 (1)
16 (1)
20 (1)
26 (1)
Gel size (cm W):
8.5 (9)
14 (3)
13 (2)
20 (2)
7.5 (2)
7 (1)
23 (1)
5.5 (1)
26 (1)
Glass size (cm H):
10 (9)
8 (9)
14.5 (7)
16 (7)
19 (7)
22 (7)
28 (7)
Glass size (cm W):
16.5 (9)
20 (5)
16 (3)
Glass size (cm L):
20 (5)
14 (3)
14.5 (1)
16 (1)
19 (1)
22 (1)
28 (1)
Capacity (cu ft):
Capacity:
0.5 mL (1)
0.2 mL (1)
2,600 pages
of answers

FREE
2013/14 Cole-Parmer
Sourcebook
Over 200 pages of life science products from industry leading brands such as Whatman, Corning, Thermo Scientific, Thermo Scientific Nalgene, Thermo Scientific NUNC, and EMD Chemicals.
Request your copy today.

Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Genomics

Genomics

For over 50 years, Cole-Parmer has been providing expert assistance and a wide selection of products to suit your requirements, including a wide variety of products and solutions for genomic research. Whether you are looking for thermal cyclers, electrophoresis products, spectrophotometers, PCR plates, or sealing films, Cole-Parmer is sure to have the right product for all your needs. Need help finding a product? Contact our Application Specialists to help find the product right for you.

Genomics refers to the study of genomes, or the complete set of genes within an organism. Mapping of genetic material and determining the entire sequence of an organisms DNA are some primary focuses at this time. Determining the genetic make-up of an organism, along with examining the environmental conditions, are primary areas of focus for examination of the roots of disease. There are five main areas of focus within genomics including:

  • Functional Genomics — determination of gene function and interaction
  • Comparative Genomics — determination of the evolution of species by studying relationships of genomic structures and functions across different species
  • Structural Genomics — determination of the protein structure within a given organism in one, two and three dimensions
  • Epigenomics — determination of the methylation patterns, as well as imprinting and packaging of DNA
  • Pharmacogenomics — determination of the influence of genetics on drug response by correlating gene expression with the efficacy of a drug

Products for Genomic Research
  • Thermal Cyclers

  • Choose from our line of thermal cyclers from Techne and
    and Labnet for all of your Genomics requirements
  • PCR Chambers

  • Choose from UV, UV Class 100, and other PCR stations
    from names like Plas-Labs, Misonix, and Labconco
  • Additional PCR Equipment

  • Choose from chilling/heating/mixing blocks, microcentrifuges,
    spectrophotometers, and thermal mixers for Genomic research
  • PCR Accessories

  • Choose from our line of products including cryogenics, PCR plates,
    pipettors and tips, sealing films, and vacuum pumps
  • Electrophoresis Products

  • Electrophoresis equipment including horizontal and vertical systems,
    power supplies, reagents, blotting systems, and gel dryers
  • Imaging Products

  • Imaging products for genomics research include autoradiography film,
    BioDoc-It, DigiDoc-It, MultiDoc-It, and PhotoDoc-It systems
  • Electroporation Products

  • Choose from our range of electroporators, buffers, cuvettes,
    and microslides for use with genomic research and studies
  • Centrifugal Concentrators

  • Products for use with genomics including centrifugal filter
    units and concentrators from Millipore and Vivaspin
  • Cloning Products

  • Products for cloning including cloning discs for use
    with genomics research and other life sciences
  • Hybridization incubators

  • Hybridization incubators for genomics include Minidizer,
    rotisserie, and shaking styles
  • UV crosslinkers

  • Systems for UV crosslinking of DNA or RNA to membranes or gene
    mapping for creating cleavage-inhibiting thymine dimers

Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer Blue Sensitive Autoradiography Film 8 x 10 100 pk

Cole-Parmer Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk

Product Type Electrophoresis Accessories,Brand Cole-Parmer
EW-21700-03
In stock.
$100.00 USD/PACKAGE

Qty:

EW-97701-86 Item 97701-82 shown

BioDoc-It Three Door Gel Imaging System with 302/365 nm 20 x 20 cm Transilluminator; 115VAC

Product Type Electrophoresis Imaging Systems,Dimensions " W 14.25,Dimensions " H 27
EW-97701-86
Call 1-888-358-4705 for availability.
$8,453.00 USD/EACH

Qty:

EW-97701-92 Item 97701-82 shown

BioDoc-It Three Door Gel Imaging System with 254/302/365 nm 20 x 20 cm Transilluminator; 230VAC

Product Type Electrophoresis Imaging Systems,Dimensions " W 14.25,Dimensions " H 27
EW-97701-92
Call 1-888-358-4705 for availability.
$8,966.00 USD/EACH

Qty:

Representative photo only Instant Blue Stain 1 liter

Instant Blue Stain, 1 liter

Product Type Electrophoresis Accessories
EW-28570-59
In stock.
$209.00 USD/EACH

Qty:

Representative photo only Programmable Power Supply 6000 VDC 150 mA 115V

Programmable Power Supply, 6000 VDC/150 mA, 115V

Dimensions " W 12-1/4,Dimensions " H 5-7/8,Dimensions " D 10-1/4
EW-28403-88
Call 1-888-358-4705 for availability.
$3,490.00 USD/EACH

Qty:

Representative photo only Electroporation system square wave

Electroporation system, square wave

Product Type Electroporation,Description Electroporation system,Applications Electroporation
EW-02894-18
Usually ships in 7 days.
$7,800.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Electrophoresis Power Supply 3000v 120v

Thermo Scientific Electrophoresis Power Supply 3000v 120v

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-50
Call 1-888-358-4705 for availability.
$2,574.00 USD/EACH

Qty:

EW-36205-01 Eppendorf<small><sup>&reg;</sup></small> Eporator<small><sup>&reg;</sup></small> System, 100-240 VAC, 50/60Hz

Eppendorf Eporator System, 100-240 VAC, 50/60Hz

Product Type Electroporation,Description Electroporator,Applications electroporation of gram-positive and gram-negative Microorganisms, bacteria, and yeast
EW-36205-01
Usually ships in 4 days.
$2,379.00 USD/EACH

Qty:

Representative photo only Vivaspin 20 Centrifugal Concentrators with PES MWCO Membranes 10 000 Daltons Pack of 48

Vivaspin 20 Centrifugal Concentrators with PES MWCO Membranes, 10,000 Daltons, Pack of 48

Product Type Centrifugal Concentrators with PES MWCO Membranes,Qty/pk 48
EW-36224-73
Usually ships in 6 days.
$367.00 USD/PACKAGE

Qty:

Representative photo only Cole Parmer 25 watt Dual Intensity 254nm Wavelength UV Transilluminator with 20cm x 20cm Filter 230VAC
EW-97622-02
Call 1-888-358-4705 for availability.
$1,685.00 USD/EACH

Qty:

EW-97700-20 PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It Imaging System with 302/365nm 20cm x 20cm Filter Transilluminator, 230VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-20
Usually ships in 10 days.
$3,944.00 USD/EACH

Qty:

EW-28575-00 Cole-Parmer<small><sup>&reg;</sup></small> Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Cole-Parmer Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Product Type Electrophoresis Vertical Systems,Buffer solution required mL 250,No. of wells 10, 14
EW-28575-00
In stock.
$375.00 USD/EACH

Qty:

Representative photo only X RAY FILM CASSETTE STAINLESS STEEL 8X10
EW-21700-43
Usually ships in 3 days.
$305.00 USD/EACH

Qty:

Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter 230VAC
EW-97623-18
Usually ships in 10 days.
$1,764.00 USD/EACH

Qty:

EW-97700-10 PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It 50 Imaging System, 115VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-10
Usually ships in 14 days.
$2,248.00 USD/EACH

Qty:

464 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title