Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Physical Property Testing
>
Viscometers
>
Falling Ball Viscometers
> Gilmont Falling Ball Viscometer
Gilmont Falling Ball Viscometer
Ideal for routine plant quality control checks and research laboratory applications
This viscometer is extremely easy to use; first fill the tube with sample, then invert tube. Next, restore tube to nornal position, turn the knob to release the ball, and then simply measure the time of descent. The ball moves against a white background with red reference lines making it easy to read. Repeatability with good technique varies from ±0.2 to ±1.0%, depending upon time of descent. Viscometers require a 7-mL sample volume.
Viscometers have wetted materials of glass, Viton
®
, and Delrin
®
acetal. PTFE O-rings are available for use with lquids that adhere to Viton
®
; call our Applications Specialists for more information.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Gilmont Falling Ball Visometer, 2 to 20 / 10 to 100 centipoise
Product Type Falling Ball Viscometer,Brand Gilmont,Model GV-2200
EW-08702-00
In stock.
$238.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Gilmont Falling Ball Viscometer
Gilmont Falling Ball Viscometer, 20 to 200 / 100 to 1000 centipoise
Product Type Falling Ball Viscometer,Brand Gilmont,Model GV-2300
EW-08702-10
In stock.
$240.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Gilmont Falling Ball Viscometer
Gilmont Fallng Ball Viscometer, 0.2 to 2 / 1 to 10 centipoise
Product Type Falling Ball Viscometer,Brand Gilmont,Model GV-2100
EW-08701-00
In stock.
$238.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Gilmont
Falling Ball Viscometer
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Physical Property Testing
>
Viscometers
>
Falling Ball Viscometers
> Gilmont Falling Ball Viscometer
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity