Global Water Easy Sensor Display
Global Water Easy Sensor Display
  • Seamless interface with 4 to 20 mA electronic sensors
  • Large LCD readout can display sensor output in any engineering units
  • Includes battery
Global Water's Easy Sensor Display features a large LCD readout that can be configured to display 4-20 mA electronic sensor values in any engineering units. The display is compatible with all of Global Water's water level, water quality, and weather 4 to 20 mA output sensors. It is housed in a rugged, weatherproof enclosure that can be purchased for either handheld or stationary use.
The sensor display is battery powered for portable handheld use. The battery powered version is also ideal for stationary appilcations when no power is available. The display stays on for as long as the disply button is pushed.
 Product Detail
Sort
 Availability
 Price
EW-99243-30 Global Water Easy Sensor Display
Global Water Easy Sensor Display
Display LCD, values up to 1900 engineering units
EW-99243-30
Usually ships in 7 days.
$411.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title