United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Turbidity Meters and Turbidity Testing Products
>
Turbidity Benchtop Meters
> Hach 2100N Laboratory Turbidity Meters
Hach 2100N Laboratory Turbidity Meters
Patented ratio optical system ensures accurate, stable readings
Smart self-diagnostics alert you to problems including using the wrong calibration standard
Meets U.S. EPA method 180.1
Wide measuring range using three selectable readout modes0 to 4000 nephelometric turbidity units, 0 to 26,800 NEPH (Nephelos), and 0 to 980 EBC (European Brewing Convention)
Optional ratioing switches instrument from multidetector ratio mode to nonratio
Signal averaging minimizes noise and display fluctuations
Air purge system connection purges the optical compartment with dry air and minimizes condensation on the sample cell
These benchtop turbidity meters offer the most versatile and advanced instruments available for nephelometric measurement. Push-button calibration and operation eliminates manual adjustments or zeroing. The patented optical system provides accurate results over a wide range of samples and turbidity, simplifying routine laboratory work.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 230 VAC
Product Type Turbidity Meter,Meter style Benchtop,Dimensions " W 15-3/4
EW-99512-07
In stock.
$2,076.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hach 2100N Laboratory Turbidity Meters
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 110 VAC
Product Type Turbidity Meter,Meter style Benchtop,Dimensions " W 15-3/4
EW-99512-00
Usually ships in 8 days.
$2,076.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hach
2100N Laboratory Turbidity Meters
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Turbidity Meters and Turbidity Testing Products
>
Turbidity Benchtop Meters
> Hach 2100N Laboratory Turbidity Meters
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title