Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Colorimetry
>
Colorimeters
> Hach DR/870 Colorimeter
Hach DR/870 Colorimeter
Faster to run samples in series—retains the selected method until a different method is selected
Performs tests for 70 different parameters
Automatically selects the wavelength for each parameter
Waterproof housing is IP67 rated and permits on-site testing
This colorimeter combines state-of-the-art features and field readiness with ease of use. The large LCD steps you through set up and procedures and indicates program number, sample number, parameter, test results, unit of measure, and more. Use prepared reagent test kits (sold separately) to run tests quickly—colorimeter comes preprogrammed for 70 parameters routinely monitored in environmental water. Memory accomodates up to 10 user-defined programs with up to 12 data points each.
Colorimeter features automatic wavelength selection, advanced reagent blank correction, and calibration curve adjustment. Analysis can be performed with user-set or pre-set reaction times through the built-in timer. Display reads results as concentration, absorbance, and percent transmittance.
Store and recall up to 99 measurements with date, time, test parameter, program number, concentration, absorbance, % transmittance, and sample number information. Data can be downloaded to a Microsoft
®
Windows
®
compatible spreadsheet or word processing program via HachLink™ software and data transfer adapter (both sold separately).
Colorimeter features a waterproof, dustproof, and chemically resistant housing.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Hach DR/870 Colorimeter
Product Type Color/Gloss Meters,Display LCD,Waterproof Yes
EW-99573-00
In stock.
$1,024.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hach DR/870 Colorimeter
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Colorimetry
>
Colorimeters
> Hach DR/870 Colorimeter
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity