log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Furniture & Fume Hoods > Fume Hoods > Ducted Fume Hoods > Hamilton Constant Volume Bypass Fume Hoods > Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Hamilton Constant Volume Benchtop Fume Hoods Assemblies
  • Fume hood and cabinet assemblies make ordering easy
  • Systems come complete with hood, blower, work surface, and cabinets
These conveniently packaged systems take the guesswork out of ordering. Assemblies include a constant volume fume hood, a work surface, your choice of standard base or chemical storage cabinet, blower, vent kit, fillers, four service fixtures, molding, and corners. Included blowers are designed for use with 35 feet (10.7 m) of ductwork and one 90º angle; use existing ductwork or install new ductwork (not included). For information on these components, contact our Application Specialists.
Fume hoods and cabinets are made of durable powder-coated steel. Hoods and cabinets are available in 18 different colors; choose color when ordering from the chart below. The 4-ft (1.2-m) hood systems come with one cabinet; the 5-, 6-, and 8-ft (1.5-, 1.8-, and 2.4-m) hood systems come with two cabinets. All models must be wired and piped. Installation required (not included).
 Product Detail
Sort
 Availability
 Price
Representative photo only Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet Epoxy Worksurface With Cup Sink
Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet Epoxy Worksurface With Cup Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 72,Exterior Dimensions " H 96
EW-41525-20
Usually ships in 13 days.
$13,858.00 USD/EACH
Qty:
Representative photo only Complete 8foot Fume Hood System With Acid And Base Storage Cabinet Epoxy Worksurface With Cup Sink
Complete 8foot Fume Hood System With Acid And Base Storage Cabinet Epoxy Worksurface With Cup Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 96,Exterior Dimensions " H 96
EW-41525-30
Usually ships in 10 days.
$13,257.00 USD/EACH
Qty:
Representative photo only Complete 5foot Fume Hood System With Acid And Base Storage Cabinets Epoxy Worksurface With Cup Sink
Complete 5foot Fume Hood System With Acid And Base Storage Cabinets Epoxy Worksurface With Cup Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 60,Exterior Dimensions " H 96
EW-41525-10
Call 1-888-358-4705 for availability.
$10,748.00 USD/EACH
Qty:
EW-41525-00 Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksurface With Cup Sink
Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksurface With Cup Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 48,Exterior Dimensions " H 96
EW-41525-00
Call 1-888-358-4705 for availability.
$9,441.00 USD/EACH
Qty:
Representative photo only Complete 8foot Fume Hood System With 2 Base Storage Cabinet Epoxy Worksurface With No Sink
Complete 8foot Fume Hood System With 2 Base Storage Cabinet Epoxy Worksurface With No Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 96,Exterior Dimensions " H 96
EW-41525-35
Call 1-888-358-4705 for availability.
$12,635.00 USD/EACH
Qty:
Representative photo only Complete 5foot Fume Hood System With 2 Base Storage Cabinet Epoxy Worksurface With No Sink
Complete 5foot Fume Hood System With 2 Base Storage Cabinet Epoxy Worksurface With No Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 60,Exterior Dimensions " H 96
EW-41525-15
Call 1-888-358-4705 for availability.
$10,139.00 USD/EACH
Qty:
Representative photo only Complete 6foot Fume Hood System With 2 Base Storage Cabinet Epoxy Worksurface With No Sink
Complete 6foot Fume Hood System With 2 Base Storage Cabinet Epoxy Worksurface With No Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 72,Exterior Dimensions " H 96
EW-41525-25
Usually ships in 10 days.
$10,945.00 USD/EACH
Qty:
Representative photo only Complete 4foot Fume Hood System With General Base Storage Cabinet Epoxy Worksurface With No Sink
Complete 4foot Fume Hood System With General Base Storage Cabinet Epoxy Worksurface With No Sink
Product Type Ducted Fume Hood,Exterior Dimensions " W 48,Exterior Dimensions " H 96
EW-41525-05
Call 1-888-358-4705 for availability.
$8,860.00 USD/EACH
Qty:
8 results (showing 1 - 8 )
Per Page 
Home  >  Furniture & Fume Hoods > Fume Hoods > Ducted Fume Hoods > Hamilton Constant Volume Bypass Fume Hoods > Hamilton Constant Volume Benchtop Fume Hoods Assemblies
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title