Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pressure & Vacuum Instruments
>
Gauge guards, Seals, and Accessories
>
Gauge Service Tools
> Hand Jack Set
Hand Jack Set
Hand Jack Set
works on all 21Ú2'' to 12'' diameter gauges. Remove or reassemble pointers without damaging your gauge. Simply slide hand jack over pointer to the movement pinion and screw down to lift pointer. Set includes a gauge-pointer remover and driver; driver features two holes to reassemble single and duplex gauges.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Hand Jack Set
Product Type Accessory,Model 3220
EW-07359-60
In stock.
$68.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hand Jack Set
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Pressure & Vacuum Instruments
>
Gauge guards, Seals, and Accessories
>
Gauge Service Tools
> Hand Jack Set
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity