United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Personal Protective Equipment (PPE)
> Hard Hats
Hard Hats
A safety hard hat may be the single most important piece of safety laboratory equipment you can purchase. Although personal protective equipment is available for most areas of the body, protecting yourself and your staff from head injuries is paramount. Even a single incident can be life-altering or lethal. OSHA surveys state that nearly two million workers may be at risk for head injuries and should use headgear such as a hard hat or safety hard hat. From construction sites to industrial facilities to manufacturing, hard hats protect workers from flying objects, head impact, falling debris, and more. Because most serious or critical head injuries result from explosions, falls, and other traumatic events, these may be prevented by the use of appropriate head protection. OSHA has established regulations for workers who may be exposed to these types of hazards.
Securing a proper fit is necessary for full head support. Cole-Parmer offers safety hard hats with ratchet suspension for convenient size adjustment. Shock-absorbing features help to evenly cushion the head from impact. Other type of safety headgear, including face shields, helmets, and more, look for personal protective equipment. If you need help selecting a safety hard hat contact a Cole-Parmer Technical Expert today.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Willson Alpha Hard Hat, Blue
Color Blue,Compliance ANSI Z89.1-1986, Class A and B,Brand Willson
EW-81665-14
In stock.
$20.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Willson
Alpha
Hard Hats
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Personal Protective Equipment (PPE)
> Hard Hats
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title