United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Heating Equipment
>
Specialty Heaters
> Heat Exchangers
Heat Exchangers
Compact design allows maximum thermal efficiency
Use to easily heat or cool virtually any liquid
These heat exchangers are a fast, economical alternative to heating or cooling liquids without the use of a tank storage system. The compact geometric design of the tubes and baffles produces the high heat transfer in a small package. These heat exchangers are great for laboratory and pilot plant installations in the petrochemical, pharmaceutical, food, and other process industries.
Small diameter tube bundles closely packed inside a shell offer maximum thermal efficiency. Curculate any two liquids with a maximum differential temperature of 150ºF (84ºC) through the heat exchanger shell and tube. Sealless, 316 stainless steel design with VITON
®
seals ensure against leaks and corrosion.
To order the correctheat exchanger for you rapplication, consider the following factors: Pressure: as a function of temperature. Heat exchangers offer a maximum steam pressure of 125 psi.
Transfer Area: determines how fast your fluid will heat or cool. Generally, the larger the transfer area the faster your product will heat/cool.
Shell and Tube Volumes: to determine if the amount of liquid will be enough to run through the shell or tube.
Hardware: to fit heat exchanger in your system. "A" (see diagram above and table below) indicates the overall tube length. Stainless stell mounting kits (sold separately below) include all of the hardware required to properly mount on heat exchanger.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Heat exchanger, 1.43 sq ft transfer area
Product Type Sanitary Heat Exchanger,Model 00277-1
EW-36125-15
In stock.
$2,215.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Heat Exchangers
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Heating Equipment
>
Specialty Heaters
> Heat Exchangers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title