Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Ovens (196)
Furnaces (187)
Burners (33)
Brand:
Product Type:
Tapes (130)
Availability:
In Stock (277)
Capacity (Liters):
Capacity (mL):
CE Compliance:
Yes (347)
Capacity (cu ft):
0 to 10 (250)
Top plate material:
Ceramic (43)
Material:
Max speed (rpm):
Drum Type:
Metal (18)
Poly (8)
Min speed (rpm):
0 to 50 (18)
Drum Size (gal.):
55 (15)
5 (6)
Control type:
Analog (2)
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Heating Equipment

Heating Equipment

Heating Equipment

Laboratory heating equipment consists of several sub-categories designed for various laboratory heating application. Each sub-category features a specific type of product for heating. Multi-purpose block heaters are ideal for incubation and activation of cultures, enzyme reactions, blood urea nitrogen determinations, immunoassays, melting/boiling points. Burners are common in laboratories that produce a single open gas flame, which is used for heating and sterilization. Drum heaters provide a practical, efficient means of freeze protection, viscosity control and maintenance of materials at elevated temperatures with in a drum. Environmental chambers create temperature humidity extremes in chambers. Furnaces are designed for laboratory and industrial applications such as moisture drying, curing, aging and general testing. Heat guns can be used for shrinking sleeves, soldering, de-soldering, paint stripping, plastic welding, drying, curing and laminating. Heat sealers are machines used to seal products, packaging, and other thermoplastic materials using heat. Heating mantles used to apply heat to containers, as an alternative to heating baths containers may be placed in direct contact with the heating mantle without substantially increasing the risk of the glassware shattering. For help with laboratory heating equipment, contact a Cole-Parmer technical expert.
 Product Detail
Sort
 Availability
 Price
Representative photo only Mechanical Convection Oven 2 1cu ft 115VAC

Mechanical Convection Oven 2.1cu.ft; 115VAC

Product Type Mechanical Convection Oven,Capacity Liters 59.451,Capacity cu ft 2.1
EW-52402-77
In stock.
$1,900.00 USD/EACH

Qty:

EW-33959-06 Yamato Programmable Mechanical Convection Oven, 5.3 Cu Ft, 115VAC

Yamato Programmable Mechanical Convection Oven, 5.3 Cu Ft, 115VAC

Product Type Mechanical Convection Oven,Capacity Liters 150.043,Capacity cu ft 5.3
EW-33959-06
In stock.
$3,120.00 USD/EACH

Qty:

EW-05053-10 StableTemp<small><sup>&reg;</sup></small> vacuum oven, 0.7 cu ft, 120 VAC

StableTemp vacuum oven, 0.7 cu ft, 120 VAC

Product Type Vacuum Oven,Capacity Liters 19.817,Capacity cu ft 0.7
EW-05053-10
In stock.
$2,830.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Thermolyne Cast Aluminum Hot Plate 12 x 12 120 VAC UL listed

Thermo Scientific Thermolyne Cast-Aluminum Hot Plate, 12" x 12", 120 VAC, UL-listed

Top plate material Aluminum,Power VAC 120,Brand Thermolyne
EW-03450-00
In stock.
$960.00 USD/EACH

Qty:

EW-33855-30 Model 33855-10 pictured.  Actual furnace may vary slightly.
EW-33855-30
In stock.
$2,600.00 USD/EACH

Qty:

Representative photo only StableTemp Economical Gravity Convection Oven 1 7 cu ft 115 VAC

StableTemp Economical Gravity Convection Oven, 1.7 cu ft, 115 VAC

Product Type Gravity Convection Oven,Capacity Liters 42.465,Capacity cu ft 1.5
EW-52120-02
Usually ships in 14 days.
$845.00 USD/EACH

Qty:

EW-37710-00 Controlled Environmental Chamber, 115 VAC, 60z

Controlled Environmental Chamber, 115 VAC, 60z

Display two 3 1/2-digit LCDs, 0.63"H,Output Humidity: 0 to 0.5V (over 0 to 100% RH span)
Temperature: 1 mV/°C or 1.8°F,Model 5518-115V
EW-37710-00
Call 1-888-358-4705 for availability.
$10,920.00 USD/EACH

Qty:

Representative photo only StableTemp furnace with programmable control 2592 cu in 208 240 VAC

StableTemp furnace with programmable control, 2592 cu in, 208/240 VAC

Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1100
EW-33858-80
Call 1-888-358-4705 for availability.
$4,145.00 USD/EACH

Qty:

Representative photo only Furnaces with three stage programmable control 565 cu in overall dimensions 15 13 16 x 16 x 17 1800 watts 200 250V

Furnaces with three-stage programmable control; 565 cu in; overall dimensions; 15 13/16" x 16" x 17"; 1800 watts; 200-250V

Product Type Muffle Furnaces,Minimum temperature º C 50,Maximum temperature º C 1100
EW-33855-35
In stock.
$2,575.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Heratherm Advanced Protocol 25 8 cuft Mechanical Convection Oven with Adjustable Fan Speed 208 240V

Thermo Scientific Heratherm Advanced Protocol 25.8 cuft Mechanical Convection Oven with Adjustable Fan Speed, 208-240V

Product Type Mechanical Convection Oven,Capacity Liters 731,Capacity cu ft 25.8
EW-38800-55
Usually ships in 6 days.
$8,100.00 USD/EACH

Qty:

Representative photo only StableTemp Ceramic Hot Plate 10 x 10 120 VAC

StableTemp Ceramic Hot Plate, 10" x 10", 120 VAC

Top plate material Ceramic,Power VAC 120,Brand StableTemp
EW-03405-30
In stock.
$415.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace 127 5 cu in 120V

Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace; 127.5 cu in; 120V

Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1100
EW-33900-10
In stock.
$1,940.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Lindberg Blue M Mini Mite Tube Furnace with 16 segment controller 120 VAC

Thermo Scientific Lindberg Blue M Mini-Mite Tube Furnace with 16-segment controller, 120 VAC

Product Type Tube Furnace,Temperature range (º C) low 100,Temperature range (º C) high 1100
EW-33850-00
Usually ships in 8 days.
$2,700.00 USD/EACH

Qty:

Representative photo only Echotherm Hot Plate Programmable Aluminum 12 X12 115V

Echotherm Hot Plate, Programmable Aluminum, 12"X12", 115V

Top plate material Aluminum,Power VAC 115,CE Compliance Yes
EW-51809-03
In stock.
$1,525.00 USD/EACH

Qty:

Representative photo only Vacuum Oven 2 3CU FT 120VAC

Vacuum Oven 2.3CU.FT; 120VAC

Product Type Mechanical Convection Oven,Capacity Liters 56.62,Capacity cu ft 2
EW-52402-96
In stock.
$4,050.00 USD/EACH

Qty:

1587 results (showing 1 - 15 )
Per Page 
Home  >  Heating Equipment

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title